Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5956336-5956563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL15RA
Entrez Gene ID
3601
Gene Name
interleukin 15 receptor subunit alpha
Gene Aliases
CD215
UniGene
Hs.445124
Gene Chromosome Location
Chr.10: 5948897-5978741 on Build GRCh38
Transcripts
NM_001243539.1 NM_001256765.1 NM_002189.3 NM_172200.2 NR_046362.1 XM_011519461.2 XM_011519462.2 XM_011519463.2 XM_011519464.2 XM_011519465.1 XM_011519466.2 XM_011519467.2 XM_011519468.1 XM_011519469.1 XM_011519470.2 XM_011519471.2 XM_011519472.2 XM_011519474.2 XM_011519475.2 XM_011519476.2 XM_011519477.2 XM_017016195.1 XM_017016196.1 XM_017016197.1 XM_017016198.1 XM_017016200.1 XM_017016201.1 XM_017016202.1 XM_017016203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6129783 c.665C>G p.S222C Chr.10: 5956406-5956406 on Build GRCh38
6684893 c.629C>T p.T210M Chr.10: 5956442-5956442 on Build GRCh38
3686795 c.677G>A p.C226Y Chr.10: 5956394-5956394 on Build GRCh38
6129784 c.665C>G p.S222C Chr.10: 5956406-5956406 on Build GRCh38
6684894 c.629C>T p.T210M Chr.10: 5956442-5956442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138544981 Chr.10: 5956493-5956493 on Build GRCh38
rs970703100 Chr.10: 5956443-5956443 on Build GRCh38
rs771511197 Chr.10: 5956471-5956471 on Build GRCh38
rs762796929 Chr.10: 5956414-5956414 on Build GRCh38
rs774028981 Chr.10: 5956462-5956462 on Build GRCh38
rs150111327 Chr.10: 5956473-5956473 on Build GRCh38
rs779123096 Chr.10: 5956371-5956371 on Build GRCh38
rs747508408 Chr.10: 5956470-5956470 on Build GRCh38
rs767438057 Chr.10: 5956502-5956502 on Build GRCh38
rs192775490 Chr.10: 5956382-5956382 on Build GRCh38
rs770899865 Chr.10: 5956395-5956395 on Build GRCh38
rs372279024 Chr.10: 5956417-5956417 on Build GRCh38
rs142602826 Chr.10: 5956442-5956442 on Build GRCh38
rs535381902 Chr.10: 5956526-5956526 on Build GRCh38
rs1037120788 Chr.10: 5956537-5956537 on Build GRCh38
rs758703676 Chr.10: 5956460-5956460 on Build GRCh38
rs750355312 Chr.10: 5956505-5956505 on Build GRCh38
rs774358512 Chr.10: 5956494-5956494 on Build GRCh38
rs1015245805 Chr.10: 5956532-5956532 on Build GRCh38
rs749995392 Chr.10: 5956433-5956433 on Build GRCh38
rs376266317 Chr.10: 5956441-5956441 on Build GRCh38
rs567583580 Chr.10: 5956368-5956368 on Build GRCh38
rs745905143 Chr.10: 5956369-5956369 on Build GRCh38
rs765547329 Chr.10: 5956412-5956412 on Build GRCh38
rs755599023 Chr.10: 5956439-5956439 on Build GRCh38
rs775877874 Chr.10: 5956413-5956413 on Build GRCh38
rs780177636 Chr.10: 5956420-5956420 on Build GRCh38
rs775791982 Chr.10: 5956487-5956487 on Build GRCh38
rs769796845 Chr.10: 5956474-5956474 on Build GRCh38
rs766935730 Chr.10: 5956432-5956432 on Build GRCh38
rs763286257 Chr.10: 5956491-5956491 on Build GRCh38
rs747528437 Chr.10: 5956393-5956393 on Build GRCh38
rs761629961 Chr.10: 5956498-5956498 on Build GRCh38
rs772052508 Chr.10: 5956397-5956397 on Build GRCh38
rs759647467 Chr.10: 5956406-5956406 on Build GRCh38
rs773640996 Chr.10: 5956389-5956389 on Build GRCh38
rs8177718 Chr.10: 5956426-5956426 on Build GRCh38
rs748458825 Chr.10: 5956373-5956373 on Build GRCh38
rs887627310 Chr.10: 5956509-5956509 on Build GRCh38

View Full Product Details