Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42233175-42233424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSK3A
Entrez Gene ID
2931
Gene Name
glycogen synthase kinase 3 alpha
Gene Aliases
-
UniGene
Hs.466828
Gene Chromosome Location
Chr.19: 42230186-42243330 on Build GRCh38
Transcripts
NM_019884.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7323577 c.1022G>A p.R341Q Chr.19: 42233186-42233186 on Build GRCh38
1293128 c.983A>G p.Q328R Chr.19: 42233305-42233305 on Build GRCh38
5449292 c.931G>A p.A311T Chr.19: 42233357-42233357 on Build GRCh38
1394065 c.1010delG p.G337fs*11 Chr.19: 42233198-42233198 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760170493 Chr.19: 42233217-42233217 on Build GRCh38
rs771203065 Chr.19: 42233214-42233214 on Build GRCh38
rs776011715 Chr.19: 42233394-42233394 on Build GRCh38
rs762576668 Chr.19: 42233260-42233260 on Build GRCh38
rs774561261 Chr.19: 42233215-42233215 on Build GRCh38
rs1001623807 Chr.19: 42233251-42233251 on Build GRCh38
rs986304454 Chr.19: 42233255-42233255 on Build GRCh38
rs751035995 Chr.19: 42233273-42233273 on Build GRCh38
rs866283999 Chr.19: 42233357-42233357 on Build GRCh38
rs767906745 Chr.19: 42233262-42233266 on Build GRCh38
rs374265274 Chr.19: 42233403-42233403 on Build GRCh38
rs777426805 Chr.19: 42233330-42233330 on Build GRCh38
rs761056625 Chr.19: 42233249-42233249 on Build GRCh38
rs867510755 Chr.19: 42233354-42233354 on Build GRCh38
rs745905801 Chr.19: 42233390-42233390 on Build GRCh38
rs376641475 Chr.19: 42233307-42233307 on Build GRCh38
rs769118918 Chr.19: 42233405-42233405 on Build GRCh38
rs868320301 Chr.19: 42233201-42233201 on Build GRCh38
rs369820952 Chr.19: 42233334-42233334 on Build GRCh38
rs549379874 Chr.19: 42233276-42233276 on Build GRCh38
rs771258407 Chr.19: 42233345-42233345 on Build GRCh38
rs749580672 Chr.19: 42233333-42233333 on Build GRCh38
rs374812848 Chr.19: 42233259-42233259 on Build GRCh38
rs768110568 Chr.19: 42233221-42233221 on Build GRCh38
rs775750857 Chr.19: 42233239-42233239 on Build GRCh38
rs371248986 Chr.19: 42233224-42233224 on Build GRCh38
rs779175432 Chr.19: 42233377-42233377 on Build GRCh38
rs764814731 Chr.19: 42233256-42233256 on Build GRCh38
rs369193412 Chr.19: 42233271-42233271 on Build GRCh38
rs772480859 Chr.19: 42233391-42233391 on Build GRCh38
rs781132710 Chr.19: 42233277-42233277 on Build GRCh38

View Full Product Details