Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62200764-62200989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTG2
Entrez Gene ID
26164
Gene Name
mitochondrial ribosome associated GTPase 2
Gene Aliases
GTPBP5, ObgH1, dJ1005F21.2
UniGene
Hs.340636
Gene Chromosome Location
Chr.20: 62183025-62203568 on Build GRCh38
Transcripts
NM_015666.3 XM_005260393.1 XM_006723774.3 XM_011528776.1 XM_017027805.1 XM_017027806.1 XM_017027807.1 XM_017027808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6716333 c.1091G>A p.G364E Chr.20: 62200947-62200947 on Build GRCh38
6716332 c.951G>A p.P317P Chr.20: 62200807-62200807 on Build GRCh38
3738000 c.954G>A p.W318* Chr.20: 62200810-62200810 on Build GRCh38
1412898 c.1020C>T p.H340H Chr.20: 62200876-62200876 on Build GRCh38
1412897 c.1010C>T p.A337V Chr.20: 62200866-62200866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774733659 Chr.20: 62200885-62200885 on Build GRCh38
rs146666461 Chr.20: 62200883-62200883 on Build GRCh38
rs749577136 Chr.20: 62200811-62200811 on Build GRCh38
rs201754148 Chr.20: 62200945-62200945 on Build GRCh38
rs761463431 Chr.20: 62200951-62200951 on Build GRCh38
rs201219598 Chr.20: 62200845-62200845 on Build GRCh38
rs926391524 Chr.20: 62200817-62200817 on Build GRCh38
rs1972 Chr.20: 62200876-62200876 on Build GRCh38
rs917467551 Chr.20: 62200849-62200849 on Build GRCh38
rs749452795 Chr.20: 62200882-62200882 on Build GRCh38
rs777760321 Chr.20: 62200968-62200968 on Build GRCh38
rs377220160 Chr.20: 62200969-62200969 on Build GRCh38
rs778978065 Chr.20: 62200972-62200972 on Build GRCh38
rs149937939 Chr.20: 62200844-62200844 on Build GRCh38
rs1035597608 Chr.20: 62200858-62200858 on Build GRCh38
rs200284014 Chr.20: 62200833-62200833 on Build GRCh38
rs374728652 Chr.20: 62200944-62200944 on Build GRCh38
rs997996464 Chr.20: 62200920-62200920 on Build GRCh38
rs781266075 Chr.20: 62200786-62200786 on Build GRCh38
rs771919472 Chr.20: 62200783-62200783 on Build GRCh38
rs760243817 Chr.20: 62200846-62200846 on Build GRCh38
rs758096165 Chr.20: 62200959-62200959 on Build GRCh38
rs148648036 Chr.20: 62200867-62200867 on Build GRCh38
rs776112821 Chr.20: 62200934-62200934 on Build GRCh38
rs772532920 Chr.20: 62200918-62200918 on Build GRCh38
rs763740889 Chr.20: 62200857-62200857 on Build GRCh38
rs753539075 Chr.20: 62200863-62200863 on Build GRCh38
rs35693261 Chr.20: 62200866-62200866 on Build GRCh38
rs748258538 Chr.20: 62200788-62200788 on Build GRCh38
rs777886666 Chr.20: 62200880-62200880 on Build GRCh38
rs912844200 Chr.20: 62200960-62200960 on Build GRCh38
rs141396472 Chr.20: 62200935-62200935 on Build GRCh38
rs774589231 Chr.20: 62200822-62200822 on Build GRCh38
rs758315602 Chr.20: 62200872-62200872 on Build GRCh38
rs139683821 Chr.20: 62200807-62200807 on Build GRCh38
rs899927361 Chr.20: 62200900-62200900 on Build GRCh38
rs954228584 Chr.20: 62200889-62200889 on Build GRCh38
rs776557341 Chr.20: 62200930-62200930 on Build GRCh38
rs35312144 Chr.20: 62200810-62200810 on Build GRCh38
rs771312911 Chr.20: 62200816-62200816 on Build GRCh38
rs746237587 Chr.20: 62200886-62200886 on Build GRCh38
rs767936109 Chr.20: 62200834-62200834 on Build GRCh38
rs937503041 Chr.20: 62200836-62200836 on Build GRCh38
rs766226812 Chr.20: 62200960-62200960 on Build GRCh38
rs756921283 Chr.20: 62200864-62200864 on Build GRCh38
rs187247879 Chr.20: 62200877-62200877 on Build GRCh38
rs754921359 Chr.20: 62200963-62200963 on Build GRCh38
rs769909865 Chr.20: 62200806-62200806 on Build GRCh38
rs764831103 Chr.20: 62200955-62200955 on Build GRCh38
rs756182304 Chr.20: 62200879-62200879 on Build GRCh38
rs143916641 Chr.20: 62200971-62200971 on Build GRCh38
rs750185399 Chr.20: 62200958-62200958 on Build GRCh38

View Full Product Details