Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26890835-26891086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH3
Entrez Gene ID
63906
Gene Name
G-patch domain containing 3
Gene Aliases
GPATC3
UniGene
Hs.10903
Gene Chromosome Location
Chr.1: 26890488-26900471 on Build GRCh38
Transcripts
NM_022078.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5380527 c.1552T>A p.S518T Chr.1: 26891036-26891036 on Build GRCh38
7155976 c.1565C>T p.S522L Chr.1: 26891023-26891023 on Build GRCh38
3804828 c.1541C>A p.S514Y Chr.1: 26891047-26891047 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778852420 Chr.1: 26890872-26890872 on Build GRCh38
rs373564628 Chr.1: 26891002-26891002 on Build GRCh38
rs766785967 Chr.1: 26891026-26891026 on Build GRCh38
rs368590257 Chr.1: 26890932-26890932 on Build GRCh38
rs776954143 Chr.1: 26890885-26890885 on Build GRCh38
rs936459069 Chr.1: 26890943-26890943 on Build GRCh38
rs765237312 Chr.1: 26891053-26891053 on Build GRCh38
rs146703973 Chr.1: 26891052-26891052 on Build GRCh38
rs1013482022 Chr.1: 26891027-26891027 on Build GRCh38
rs372484136 Chr.1: 26890882-26890883 on Build GRCh38
rs776816476 Chr.1: 26890998-26890998 on Build GRCh38
rs747713930 Chr.1: 26891004-26891004 on Build GRCh38
rs370963008 Chr.1: 26891051-26891051 on Build GRCh38
rs779533297 Chr.1: 26890981-26890981 on Build GRCh38
rs775255134 Chr.1: 26890882-26890882 on Build GRCh38
rs781340687 Chr.1: 26890965-26890965 on Build GRCh38
rs192893502 Chr.1: 26890911-26890911 on Build GRCh38
rs556949627 Chr.1: 26890867-26890867 on Build GRCh38
rs184205112 Chr.1: 26890976-26890976 on Build GRCh38
rs765800672 Chr.1: 26890893-26890893 on Build GRCh38
rs754960489 Chr.1: 26890963-26890963 on Build GRCh38
rs377515706 Chr.1: 26891021-26891021 on Build GRCh38
rs200641480 Chr.1: 26891025-26891025 on Build GRCh38
rs764412020 Chr.1: 26890990-26890990 on Build GRCh38
rs544759803 Chr.1: 26890974-26890974 on Build GRCh38
rs773746831 Chr.1: 26891019-26891019 on Build GRCh38
rs760738069 Chr.1: 26890867-26890868 on Build GRCh38
rs770788217 Chr.1: 26890870-26890870 on Build GRCh38
rs746978696 Chr.1: 26891031-26891031 on Build GRCh38
rs762219194 Chr.1: 26891058-26891059 on Build GRCh38
rs376902197 Chr.1: 26891007-26891007 on Build GRCh38
rs374416440 Chr.1: 26890895-26890895 on Build GRCh38
rs377480001 Chr.1: 26890884-26890884 on Build GRCh38
rs767702505 Chr.1: 26891045-26891045 on Build GRCh38
rs573989936 Chr.1: 26890886-26890886 on Build GRCh38
rs945456148 Chr.1: 26890902-26890902 on Build GRCh38
rs536215879 Chr.1: 26890905-26890905 on Build GRCh38
rs762312132 Chr.1: 26890889-26890889 on Build GRCh38
rs759813924 Chr.1: 26891043-26891043 on Build GRCh38
rs771834844 Chr.1: 26890879-26890879 on Build GRCh38
rs745721698 Chr.1: 26890875-26890875 on Build GRCh38
rs138696335 Chr.1: 26891038-26891038 on Build GRCh38
rs375769609 Chr.1: 26890980-26890980 on Build GRCh38
rs572901412 Chr.1: 26890962-26890962 on Build GRCh38
rs772975044 Chr.1: 26890966-26890966 on Build GRCh38
rs917085849 Chr.1: 26890929-26890929 on Build GRCh38
rs749274326 Chr.1: 26890868-26890868 on Build GRCh38
rs763354905 Chr.1: 26890896-26890896 on Build GRCh38
rs757259598 Chr.1: 26890968-26890968 on Build GRCh38
rs776199914 Chr.1: 26891003-26891003 on Build GRCh38
rs746698081 Chr.1: 26890991-26890991 on Build GRCh38

View Full Product Details