Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92249703-92249964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS
Entrez Gene ID
3376
Gene Name
isoleucyl-tRNA synthetase
Gene Aliases
IARS1, ILERS, ILRS, IRS, PRO0785
UniGene
Hs.445403
Gene Chromosome Location
Chr.9: 92210207-92293756 on Build GRCh38
Transcripts
NM_002161.5 NM_013417.3 NR_073446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5926662 c.2536C>T p.P846S Chr.9: 92249938-92249938 on Build GRCh38
6208653 c.2585A>T p.D862V Chr.9: 92249889-92249889 on Build GRCh38
487660 c.2557A>T p.I853F Chr.9: 92249917-92249917 on Build GRCh38
311904 c.2610C>G p.I870M Chr.9: 92249864-92249864 on Build GRCh38
487661 c.2555T>C p.V852A Chr.9: 92249919-92249919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533775614 Chr.9: 92249765-92249765 on Build GRCh38
rs769603552 Chr.9: 92249887-92249887 on Build GRCh38
rs754567807 Chr.9: 92249808-92249808 on Build GRCh38
rs749495793 Chr.9: 92249854-92249854 on Build GRCh38
rs966175198 Chr.9: 92249832-92249832 on Build GRCh38
rs748154039 Chr.9: 92249881-92249881 on Build GRCh38
rs778064736 Chr.9: 92249851-92249851 on Build GRCh38
rs776350346 Chr.9: 92249822-92249822 on Build GRCh38
rs757465898 Chr.9: 92249865-92249865 on Build GRCh38
rs763167401 Chr.9: 92249915-92249915 on Build GRCh38
rs187938830 Chr.9: 92249781-92249781 on Build GRCh38
rs1039213837 Chr.9: 92249746-92249746 on Build GRCh38
rs900215067 Chr.9: 92249785-92249785 on Build GRCh38
rs999093095 Chr.9: 92249866-92249866 on Build GRCh38
rs572669213 Chr.9: 92249743-92249743 on Build GRCh38
rs573844945 Chr.9: 92249782-92249782 on Build GRCh38
rs756532539 Chr.9: 92249828-92249828 on Build GRCh38
rs562858838 Chr.9: 92249923-92249923 on Build GRCh38
rs753267867 Chr.9: 92249824-92249824 on Build GRCh38
rs768072045 Chr.9: 92249823-92249823 on Build GRCh38
rs41280223 Chr.9: 92249786-92249786 on Build GRCh38
rs779214663 Chr.9: 92249868-92249868 on Build GRCh38
rs932022371 Chr.9: 92249887-92249888 on Build GRCh38
rs776539941 Chr.9: 92249880-92249880 on Build GRCh38
rs908043078 Chr.9: 92249831-92249831 on Build GRCh38
rs772141867 Chr.9: 92249873-92249873 on Build GRCh38
rs376888710 Chr.9: 92249843-92249843 on Build GRCh38
rs769637165 Chr.9: 92249814-92249819 on Build GRCh38
rs369345893 Chr.9: 92249924-92249924 on Build GRCh38
rs983187027 Chr.9: 92249812-92249812 on Build GRCh38

View Full Product Details