Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60043493-60043764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR6
Entrez Gene ID
63897
Gene Name
HEAT repeat containing 6
Gene Aliases
ABC1
UniGene
Hs.221899
Gene Chromosome Location
Chr.17: 60041363-60078931 on Build GRCh38
Transcripts
NM_022070.4 XM_005257570.3 XM_011525110.2 XM_011525111.2 XM_017024936.1 XM_017024937.1 XM_017024938.1 XM_017024939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6217386 c.3442G>T p.D1148Y Chr.17: 60043667-60043667 on Build GRCh38
6901176 c.3526C>T p.P1176S Chr.17: 60043583-60043583 on Build GRCh38
4414749 c.3434C>A p.P1145Q Chr.17: 60043675-60043675 on Build GRCh38
7242118 c.3528A>T p.P1176P Chr.17: 60043581-60043581 on Build GRCh38
218526 c.3430G>C p.A1144P Chr.17: 60043679-60043679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762569216 Chr.17: 60043575-60043575 on Build GRCh38
rs767017193 Chr.17: 60043679-60043679 on Build GRCh38
rs757931443 Chr.17: 60043519-60043519 on Build GRCh38
rs769144742 Chr.17: 60043745-60043745 on Build GRCh38
rs757059343 Chr.17: 60043710-60043710 on Build GRCh38
rs746062275 Chr.17: 60043731-60043731 on Build GRCh38
rs779279891 Chr.17: 60043722-60043722 on Build GRCh38
rs772444281 Chr.17: 60043558-60043558 on Build GRCh38
rs777662012 Chr.17: 60043595-60043595 on Build GRCh38
rs748503657 Chr.17: 60043673-60043673 on Build GRCh38
rs571183486 Chr.17: 60043528-60043528 on Build GRCh38
rs755892944 Chr.17: 60043594-60043594 on Build GRCh38
rs781111870 Chr.17: 60043531-60043531 on Build GRCh38
rs778248572 Chr.17: 60043592-60043593 on Build GRCh38
rs147940060 Chr.17: 60043570-60043570 on Build GRCh38
rs562782641 Chr.17: 60043572-60043572 on Build GRCh38
rs141770217 Chr.17: 60043588-60043588 on Build GRCh38
rs558403592 Chr.17: 60043593-60043593 on Build GRCh38
rs993879794 Chr.17: 60043665-60043665 on Build GRCh38
rs750094327 Chr.17: 60043574-60043574 on Build GRCh38
rs768990601 Chr.17: 60043604-60043605 on Build GRCh38
rs756056393 Chr.17: 60043687-60043687 on Build GRCh38
rs895721139 Chr.17: 60043662-60043663 on Build GRCh38
rs201811006 Chr.17: 60043529-60043529 on Build GRCh38
rs761191743 Chr.17: 60043642-60043642 on Build GRCh38
rs115876236 Chr.17: 60043662-60043662 on Build GRCh38
rs200810311 Chr.17: 60043536-60043536 on Build GRCh38
rs533285312 Chr.17: 60043606-60043606 on Build GRCh38
rs5821289 Chr.17: 60043592-60043592 on Build GRCh38
rs751067048 Chr.17: 60043582-60043582 on Build GRCh38
rs375867425 Chr.17: 60043545-60043545 on Build GRCh38
rs138450019 Chr.17: 60043740-60043740 on Build GRCh38
rs776947371 Chr.17: 60043652-60043652 on Build GRCh38
rs770179982 Chr.17: 60043718-60043718 on Build GRCh38
rs752599831 Chr.17: 60043683-60043683 on Build GRCh38
rs370995532 Chr.17: 60043517-60043517 on Build GRCh38
rs774552842 Chr.17: 60043549-60043549 on Build GRCh38
rs780157364 Chr.17: 60043739-60043739 on Build GRCh38
rs745390730 Chr.17: 60043593-60043594 on Build GRCh38
rs149927682 Chr.17: 60043619-60043619 on Build GRCh38
rs760952437 Chr.17: 60043571-60043571 on Build GRCh38
rs779189940 Chr.17: 60043608-60043608 on Build GRCh38
rs749482636 Chr.17: 60043537-60043537 on Build GRCh38
rs755002788 Chr.17: 60043587-60043587 on Build GRCh38
rs749418877 Chr.17: 60043655-60043655 on Build GRCh38
rs751156949 Chr.17: 60043666-60043666 on Build GRCh38
rs777184497 Chr.17: 60043616-60043616 on Build GRCh38
rs765911274 Chr.17: 60043579-60043579 on Build GRCh38
rs577133638 Chr.17: 60043530-60043530 on Build GRCh38
rs756958988 Chr.17: 60043589-60043591 on Build GRCh38
rs538273826 Chr.17: 60043580-60043580 on Build GRCh38
rs371805947 Chr.17: 60043678-60043678 on Build GRCh38
rs1047165673 Chr.17: 60043673-60043673 on Build GRCh38
rs145634926 Chr.17: 60043591-60043591 on Build GRCh38
rs368238079 Chr.17: 60043625-60043625 on Build GRCh38
rs758429792 Chr.17: 60043737-60043737 on Build GRCh38
rs772122728 Chr.17: 60043620-60043620 on Build GRCh38
rs753683192 Chr.17: 60043709-60043709 on Build GRCh38
rs140665566 Chr.17: 60043647-60043647 on Build GRCh38
rs749572665 Chr.17: 60043605-60043605 on Build GRCh38
rs116244621 Chr.17: 60043697-60043697 on Build GRCh38

View Full Product Details