Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68303825-68304058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA11
Entrez Gene ID
22801
Gene Name
integrin subunit alpha 11
Gene Aliases
HsT18964
UniGene
Hs.436416
Gene Chromosome Location
Chr.15: 68297433-68432312 on Build GRCh38
Transcripts
NM_001004439.1 XM_005254228.3 XM_011521363.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7223244 c.3419T>G p.V1140G Chr.15: 68303848-68303848 on Build GRCh38
4602390 c.3418G>T p.V1140F Chr.15: 68303849-68303849 on Build GRCh38
6303627 c.3411C>A p.D1137E Chr.15: 68303856-68303856 on Build GRCh38
5812925 c.3412T>A p.W1138R Chr.15: 68303855-68303855 on Build GRCh38
4489038 c.3421C>T p.P1141S Chr.15: 68303846-68303846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372051800 Chr.15: 68303896-68303896 on Build GRCh38
rs952227579 Chr.15: 68303942-68303942 on Build GRCh38
rs201171435 Chr.15: 68303865-68303865 on Build GRCh38
rs367755317 Chr.15: 68303866-68303866 on Build GRCh38
rs986057366 Chr.15: 68303963-68303963 on Build GRCh38
rs200364693 Chr.15: 68303883-68303883 on Build GRCh38
rs1035056399 Chr.15: 68303870-68303870 on Build GRCh38
rs79481898 Chr.15: 68303848-68303848 on Build GRCh38
rs760816337 Chr.15: 68303856-68303856 on Build GRCh38
rs775809251 Chr.15: 68303896-68303897 on Build GRCh38
rs760913611 Chr.15: 68303905-68303905 on Build GRCh38
rs200383813 Chr.15: 68303933-68303933 on Build GRCh38
rs369769254 Chr.15: 68303897-68303897 on Build GRCh38
rs557666707 Chr.15: 68304007-68304007 on Build GRCh38
rs975709374 Chr.15: 68303931-68303931 on Build GRCh38
rs757056428 Chr.15: 68303880-68303880 on Build GRCh38
rs985472966 Chr.15: 68303951-68303951 on Build GRCh38
rs936499071 Chr.15: 68303998-68303998 on Build GRCh38
rs771208316 Chr.15: 68303925-68303925 on Build GRCh38
rs772230129 Chr.15: 68303886-68303887 on Build GRCh38
rs939685414 Chr.15: 68303983-68303983 on Build GRCh38
rs746568016 Chr.15: 68303898-68303898 on Build GRCh38
rs754476863 Chr.15: 68303901-68303901 on Build GRCh38
rs565489893 Chr.15: 68303904-68303904 on Build GRCh38
rs749748531 Chr.15: 68303912-68303912 on Build GRCh38
rs774820701 Chr.15: 68303928-68303928 on Build GRCh38
rs574276907 Chr.15: 68304011-68304011 on Build GRCh38
rs557420599 Chr.15: 68303936-68303936 on Build GRCh38
rs142182120 Chr.15: 68303960-68303960 on Build GRCh38
rs925218731 Chr.15: 68303977-68303977 on Build GRCh38
rs535707272 Chr.15: 68303859-68303859 on Build GRCh38
rs778203666 Chr.15: 68303905-68303905 on Build GRCh38
rs536618289 Chr.15: 68304005-68304005 on Build GRCh38
rs547940090 Chr.15: 68303849-68303849 on Build GRCh38

View Full Product Details