Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75727615-75727840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB4
Entrez Gene ID
3691
Gene Name
integrin subunit beta 4
Gene Aliases
CD104, GP150
UniGene
Hs.632226
Gene Chromosome Location
Chr.17: 75721328-75757819 on Build GRCh38
Transcripts
NM_000213.4 NM_001005619.1 NM_001005731.2 NM_001321123.1 XM_005257309.2 XM_005257311.4 XM_006721866.3 XM_006721867.3 XM_006721868.3 XM_006721870.3 XM_011524751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6423105 c.413T>C p.F138S Chr.17: 75727799-75727799 on Build GRCh38
4693388 c.348G>A p.R116R Chr.17: 75727734-75727734 on Build GRCh38
6895597 c.402C>T p.I134I Chr.17: 75727788-75727788 on Build GRCh38
1610776 c.422C>T p.S141F Chr.17: 75727808-75727808 on Build GRCh38
5102371 c.326G>A p.R109H Chr.17: 75727712-75727712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745742570 Chr.17: 75727715-75727715 on Build GRCh38
rs1049605905 Chr.17: 75727712-75727712 on Build GRCh38
rs752981908 Chr.17: 75727754-75727754 on Build GRCh38
rs773439891 Chr.17: 75727731-75727731 on Build GRCh38
rs770380318 Chr.17: 75727711-75727711 on Build GRCh38
rs146269824 Chr.17: 75727732-75727732 on Build GRCh38
rs143114124 Chr.17: 75727679-75727679 on Build GRCh38
rs753473264 Chr.17: 75727782-75727782 on Build GRCh38
rs756909804 Chr.17: 75727786-75727786 on Build GRCh38
rs113072439 Chr.17: 75727778-75727778 on Build GRCh38
rs778409425 Chr.17: 75727714-75727714 on Build GRCh38
rs769872605 Chr.17: 75727722-75727722 on Build GRCh38
rs141930616 Chr.17: 75727774-75727774 on Build GRCh38
rs146338793 Chr.17: 75727793-75727793 on Build GRCh38
rs775271245 Chr.17: 75727676-75727676 on Build GRCh38
rs755554730 Chr.17: 75727693-75727693 on Build GRCh38
rs764405265 Chr.17: 75727696-75727696 on Build GRCh38
rs1033883152 Chr.17: 75727797-75727797 on Build GRCh38
rs760570610 Chr.17: 75727719-75727719 on Build GRCh38
rs752061535 Chr.17: 75727692-75727692 on Build GRCh38
rs914024305 Chr.17: 75727723-75727723 on Build GRCh38
rs760115004 Chr.17: 75727744-75727744 on Build GRCh38
rs769490746 Chr.17: 75727644-75727644 on Build GRCh38
rs748885598 Chr.17: 75727659-75727659 on Build GRCh38
rs748925101 Chr.17: 75727706-75727706 on Build GRCh38
rs141385926 Chr.17: 75727717-75727717 on Build GRCh38
rs763268075 Chr.17: 75727695-75727695 on Build GRCh38
rs750443550 Chr.17: 75727685-75727685 on Build GRCh38
rs369326405 Chr.17: 75727675-75727675 on Build GRCh38
rs200008946 Chr.17: 75727718-75727718 on Build GRCh38
rs373041337 Chr.17: 75727671-75727671 on Build GRCh38
rs770744295 Chr.17: 75727661-75727661 on Build GRCh38
rs369576655 Chr.17: 75727637-75727637 on Build GRCh38
rs761940483 Chr.17: 75727678-75727678 on Build GRCh38
rs768196342 Chr.17: 75727745-75727745 on Build GRCh38
rs777522343 Chr.17: 75727705-75727705 on Build GRCh38
rs146609350 Chr.17: 75727668-75727668 on Build GRCh38
rs781705263 Chr.17: 75727633-75727633 on Build GRCh38
rs993763228 Chr.17: 75727771-75727771 on Build GRCh38
rs997150898 Chr.17: 75727665-75727665 on Build GRCh38
rs568835164 Chr.17: 75727698-75727698 on Build GRCh38
rs140040189 Chr.17: 75727773-75727773 on Build GRCh38
rs777144015 Chr.17: 75727645-75727645 on Build GRCh38
rs751692203 Chr.17: 75727743-75727743 on Build GRCh38

View Full Product Details