Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123606580-123606851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1B
Entrez Gene ID
57476
Gene Name
GRAM domain containing 1B
Gene Aliases
-
UniGene
Hs.144725
Gene Chromosome Location
Chr.11: 123358299-123627771 on Build GRCh38
Transcripts
NM_001286563.1 NM_001286564.1 NM_020716.2 XM_005271619.4 XM_005271621.4 XM_005271623.4 XM_005271624.4 XM_006718892.3 XM_011542926.2 XM_011542929.2 XM_011542930.2 XM_011542931.2 XM_011542933.1 XM_017018034.1 XM_017018035.1 XM_017018036.1 XM_017018037.1 XM_017018038.1 XM_017018039.1 XM_017018040.1 XM_017018041.1 XM_017018042.1 XM_017018043.1 XM_017018044.1 XM_017018045.1 XM_017018046.1 XM_017018047.1 XM_017018048.1 XM_017018049.1 XM_017018050.1 XM_017018051.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5553250 c.916G>A p.E306K Chr.11: 123606630-123606630 on Build GRCh38
4018807 c.1024T>G p.F342V Chr.11: 123606738-123606738 on Build GRCh38
3359061 c.1059T>A p.S353R Chr.11: 123606773-123606773 on Build GRCh38
4018809 c.1031A>G p.D344G Chr.11: 123606745-123606745 on Build GRCh38
4833634 c.1016C>T p.S339L Chr.11: 123606730-123606730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375511617 Chr.11: 123606672-123606672 on Build GRCh38
rs999104736 Chr.11: 123606755-123606755 on Build GRCh38
rs554242193 Chr.11: 123606767-123606767 on Build GRCh38
rs1029451265 Chr.11: 123606741-123606741 on Build GRCh38
rs2279519 Chr.11: 123606644-123606644 on Build GRCh38
rs781758268 Chr.11: 123606821-123606821 on Build GRCh38
rs191981781 Chr.11: 123606675-123606675 on Build GRCh38
rs149488546 Chr.11: 123606664-123606664 on Build GRCh38
rs1031717473 Chr.11: 123606822-123606822 on Build GRCh38
rs373808855 Chr.11: 123606621-123606621 on Build GRCh38
rs747374034 Chr.11: 123606603-123606603 on Build GRCh38
rs777056555 Chr.11: 123606611-123606611 on Build GRCh38
rs769142319 Chr.11: 123606605-123606605 on Build GRCh38
rs751049869 Chr.11: 123606783-123606783 on Build GRCh38
rs765931007 Chr.11: 123606782-123606782 on Build GRCh38
rs747509131 Chr.11: 123606655-123606655 on Build GRCh38
rs753149279 Chr.11: 123606828-123606828 on Build GRCh38
rs756933883 Chr.11: 123606650-123606650 on Build GRCh38
rs757410799 Chr.11: 123606818-123606819 on Build GRCh38
rs373474951 Chr.11: 123606710-123606710 on Build GRCh38
rs371265661 Chr.11: 123606687-123606687 on Build GRCh38
rs887690377 Chr.11: 123606698-123606698 on Build GRCh38
rs774236612 Chr.11: 123606700-123606700 on Build GRCh38
rs757894340 Chr.11: 123606652-123606652 on Build GRCh38
rs267602745 Chr.11: 123606666-123606666 on Build GRCh38
rs866395584 Chr.11: 123606657-123606657 on Build GRCh38
rs867480430 Chr.11: 123606789-123606789 on Build GRCh38
rs1046775434 Chr.11: 123606670-123606670 on Build GRCh38
rs770078855 Chr.11: 123606620-123606620 on Build GRCh38
rs748398230 Chr.11: 123606616-123606616 on Build GRCh38
rs760520842 Chr.11: 123606641-123606641 on Build GRCh38
rs761575476 Chr.11: 123606726-123606726 on Build GRCh38
rs776432140 Chr.11: 123606711-123606711 on Build GRCh38
rs764839388 Chr.11: 123606730-123606730 on Build GRCh38
rs771946953 Chr.11: 123606636-123606636 on Build GRCh38
rs753560680 Chr.11: 123606646-123606646 on Build GRCh38
rs369660111 Chr.11: 123606638-123606638 on Build GRCh38
rs76756402 Chr.11: 123606705-123606705 on Build GRCh38
rs377539345 Chr.11: 123606742-123606742 on Build GRCh38
rs532856454 Chr.11: 123606637-123606637 on Build GRCh38
rs759359467 Chr.11: 123606617-123606617 on Build GRCh38
rs780494102 Chr.11: 123606653-123606653 on Build GRCh38
rs771026857 Chr.11: 123606692-123606692 on Build GRCh38
rs1030348550 Chr.11: 123606625-123606625 on Build GRCh38
rs866068201 Chr.11: 123606754-123606754 on Build GRCh38
rs370508125 Chr.11: 123606704-123606704 on Build GRCh38
rs755577223 Chr.11: 123606792-123606792 on Build GRCh38
rs376872587 Chr.11: 123606684-123606684 on Build GRCh38
rs114469707 Chr.11: 123606682-123606682 on Build GRCh38
rs749953166 Chr.11: 123606651-123606651 on Build GRCh38

View Full Product Details