Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 35634151-35634376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERPUD2
Entrez Gene ID
64224
Gene Name
HERPUD family member 2
Gene Aliases
-
UniGene
Hs.643051
Gene Chromosome Location
Chr.7: 35632659-35695187 on Build GRCh38
Transcripts
NM_022373.4 XM_006715765.1 XM_011515473.1 XM_017012506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3880786 c.1013T>G p.V338G Chr.7: 35634358-35634358 on Build GRCh38
3228711 c.999C>T p.N333N Chr.7: 35634372-35634372 on Build GRCh38
6871659 c.996C>T p.P332P Chr.7: 35634375-35634375 on Build GRCh38
6871658 c.995C>T p.P332L Chr.7: 35634376-35634376 on Build GRCh38
3638402 c.1012G>A p.V338I Chr.7: 35634359-35634359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373165428 Chr.7: 35634263-35634263 on Build GRCh38
rs757355209 Chr.7: 35634290-35634290 on Build GRCh38
rs962664866 Chr.7: 35634200-35634200 on Build GRCh38
rs763287535 Chr.7: 35634273-35634273 on Build GRCh38
rs778569836 Chr.7: 35634348-35634348 on Build GRCh38
rs778970288 Chr.7: 35634293-35634293 on Build GRCh38
rs906938115 Chr.7: 35634245-35634245 on Build GRCh38
rs934088643 Chr.7: 35634257-35634257 on Build GRCh38
rs991058018 Chr.7: 35634186-35634186 on Build GRCh38
rs768340617 Chr.7: 35634345-35634345 on Build GRCh38
rs555647618 Chr.7: 35634247-35634247 on Build GRCh38
rs555155528 Chr.7: 35634202-35634202 on Build GRCh38
rs972737111 Chr.7: 35634216-35634216 on Build GRCh38
rs750348270 Chr.7: 35634311-35634311 on Build GRCh38
rs764377315 Chr.7: 35634279-35634279 on Build GRCh38
rs557289330 Chr.7: 35634344-35634344 on Build GRCh38
rs2305333 Chr.7: 35634189-35634189 on Build GRCh38
rs751738788 Chr.7: 35634281-35634281 on Build GRCh38
rs2305334 Chr.7: 35634260-35634260 on Build GRCh38
rs748465289 Chr.7: 35634221-35634221 on Build GRCh38
rs779913555 Chr.7: 35634339-35634339 on Build GRCh38
rs781444262 Chr.7: 35634298-35634298 on Build GRCh38
rs375276167 Chr.7: 35634319-35634319 on Build GRCh38
rs369496104 Chr.7: 35634261-35634261 on Build GRCh38
rs959589043 Chr.7: 35634178-35634178 on Build GRCh38

View Full Product Details