Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16153028-16153255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSH2D
Entrez Gene ID
84941
Gene Name
hematopoietic SH2 domain containing
Gene Aliases
ALX, HSH2
UniGene
Hs.631617
Gene Chromosome Location
Chr.19: 16134028-16158575 on Build GRCh38
Transcripts
NM_001291274.1 NM_032855.3 NR_111903.1 NR_111904.1 XM_011528386.2 XM_011528387.2 XM_011528388.2 XM_011528389.2 XM_011528390.1 XM_017027402.1 XM_017027403.1 XM_017027404.1

SNP Detail

SNP ID Chromosome Location
rs762887889 Chr.19: 16153061-16153061 on Build GRCh38
rs756362813 Chr.19: 16153115-16153115 on Build GRCh38
rs1870354 Chr.19: 16153136-16153136 on Build GRCh38
rs774911261 Chr.19: 16153057-16153057 on Build GRCh38
rs763563060 Chr.19: 16153196-16153196 on Build GRCh38
rs376493778 Chr.19: 16153168-16153168 on Build GRCh38
rs867048971 Chr.19: 16153174-16153174 on Build GRCh38
rs1010020319 Chr.19: 16153089-16153089 on Build GRCh38
rs758938321 Chr.19: 16153191-16153192 on Build GRCh38
rs371623069 Chr.19: 16153172-16153172 on Build GRCh38
rs745522000 Chr.19: 16153164-16153164 on Build GRCh38
rs184059390 Chr.19: 16153128-16153128 on Build GRCh38
rs767374701 Chr.19: 16153219-16153219 on Build GRCh38
rs772512381 Chr.19: 16153137-16153137 on Build GRCh38
rs767193480 Chr.19: 16153112-16153112 on Build GRCh38
rs868213276 Chr.19: 16153173-16153173 on Build GRCh38
rs374295346 Chr.19: 16153221-16153221 on Build GRCh38
rs761605157 Chr.19: 16153214-16153214 on Build GRCh38
rs188745915 Chr.19: 16153176-16153176 on Build GRCh38
rs751443761 Chr.19: 16153103-16153103 on Build GRCh38
rs983873538 Chr.19: 16153138-16153138 on Build GRCh38
rs1048229895 Chr.19: 16153107-16153107 on Build GRCh38
rs370655028 Chr.19: 16153187-16153187 on Build GRCh38
rs373382652 Chr.19: 16153158-16153158 on Build GRCh38
rs762512362 Chr.19: 16153177-16153177 on Build GRCh38
rs764073276 Chr.19: 16153098-16153098 on Build GRCh38
rs750678673 Chr.19: 16153113-16153113 on Build GRCh38
rs373172020 Chr.19: 16153197-16153197 on Build GRCh38
rs781347192 Chr.19: 16153216-16153216 on Build GRCh38
rs374950543 Chr.19: 16153129-16153129 on Build GRCh38
rs978492165 Chr.19: 16153218-16153218 on Build GRCh38
rs552366870 Chr.19: 16153109-16153109 on Build GRCh38
rs780295676 Chr.19: 16153122-16153122 on Build GRCh38
rs779518397 Chr.19: 16153134-16153134 on Build GRCh38
rs369342750 Chr.19: 16153106-16153106 on Build GRCh38

View Full Product Details