Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46835299-46835538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR2A
Entrez Gene ID
3356
Gene Name
5-hydroxytryptamine receptor 2A
Gene Aliases
5-HT2A, HTR2
UniGene
Hs.72630
Gene Chromosome Location
Chr.13: 46831542-46897076 on Build GRCh38
Transcripts
NM_000621.4 NM_001165947.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7112510 c.663C>T p.S221S Chr.13: 46835338-46835338 on Build GRCh38
7133660 c.652G>A p.E218K Chr.13: 46835349-46835349 on Build GRCh38
5842598 c.811G>A p.D271N Chr.13: 46835442-46835442 on Build GRCh38
1684368 c.925_927delAGG p.R310delR Chr.13: 46835326-46835328 on Build GRCh38
7154754 c.611C>G p.S204C Chr.13: 46835390-46835390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759251466 Chr.13: 46835341-46835341 on Build GRCh38
rs752088874 Chr.13: 46835344-46835344 on Build GRCh38
rs757334801 Chr.13: 46835442-46835442 on Build GRCh38
rs773797673 Chr.13: 46835333-46835333 on Build GRCh38
rs755187812 Chr.13: 46835372-46835372 on Build GRCh38
rs750188721 Chr.13: 46835479-46835479 on Build GRCh38
rs1041110431 Chr.13: 46835334-46835334 on Build GRCh38
rs376305063 Chr.13: 46835364-46835364 on Build GRCh38
rs757096279 Chr.13: 46835363-46835363 on Build GRCh38
rs748134081 Chr.13: 46835394-46835394 on Build GRCh38
rs763899284 Chr.13: 46835349-46835349 on Build GRCh38
rs533383790 Chr.13: 46835429-46835429 on Build GRCh38
rs1034308865 Chr.13: 46835360-46835360 on Build GRCh38
rs758390994 Chr.13: 46835509-46835509 on Build GRCh38
rs749521476 Chr.13: 46835366-46835366 on Build GRCh38
rs768848048 Chr.13: 46835324-46835324 on Build GRCh38
rs760992919 Chr.13: 46835335-46835335 on Build GRCh38
rs773566160 Chr.13: 46835407-46835407 on Build GRCh38
rs764945857 Chr.13: 46835342-46835342 on Build GRCh38
rs753008959 Chr.13: 46835488-46835488 on Build GRCh38
rs747569445 Chr.13: 46835413-46835413 on Build GRCh38
rs762338994 Chr.13: 46835430-46835430 on Build GRCh38
rs778969781 Chr.13: 46835485-46835485 on Build GRCh38
rs1006892485 Chr.13: 46835327-46835327 on Build GRCh38
rs766775131 Chr.13: 46835338-46835338 on Build GRCh38
rs771279570 Chr.13: 46835415-46835415 on Build GRCh38
rs192924120 Chr.13: 46835345-46835345 on Build GRCh38
rs563077877 Chr.13: 46835428-46835428 on Build GRCh38
rs776914593 Chr.13: 46835423-46835423 on Build GRCh38
rs140854817 Chr.13: 46835359-46835359 on Build GRCh38
rs751532511 Chr.13: 46835432-46835432 on Build GRCh38
rs779212618 Chr.13: 46835373-46835373 on Build GRCh38
rs772230929 Chr.13: 46835403-46835403 on Build GRCh38
rs755964470 Chr.13: 46835481-46835481 on Build GRCh38
rs767568760 Chr.13: 46835458-46835458 on Build GRCh38
rs758773486 Chr.13: 46835411-46835411 on Build GRCh38
rs776251555 Chr.13: 46835339-46835339 on Build GRCh38
rs1023428680 Chr.13: 46835426-46835426 on Build GRCh38
rs774363482 Chr.13: 46835325-46835325 on Build GRCh38
rs780484633 Chr.13: 46835415-46835416 on Build GRCh38
rs561538664 Chr.13: 46835332-46835332 on Build GRCh38
rs927684489 Chr.13: 46835500-46835500 on Build GRCh38

View Full Product Details