Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 13299126-13299399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISM1
Entrez Gene ID
140862
Gene Name
isthmin 1, angiogenesis inhibitor
Gene Aliases
C20orf82, ISM, Isthmin, bA149I18.1, dJ1077I2.1
UniGene
Hs.559353
Gene Chromosome Location
Chr.20: 13221771-13326734 on Build GRCh38
Transcripts
NM_080826.1 XM_017027680.1

Gene Symbol
TASP1
Entrez Gene ID
55617
Gene Name
taspase 1
Gene Aliases
C20orf13, dJ585I14.2
UniGene
Hs.348297
Gene Chromosome Location
Chr.20: 13104760-13638936 on Build GRCh38
Transcripts
NM_080826.1 XM_017027680.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6092647 c.1308C>A p.A436A Chr.20: 13299372-13299372 on Build GRCh38
6159210 c.1199C>G p.A400G Chr.20: 13299263-13299263 on Build GRCh38
3840428 c.1126G>A p.E376K Chr.20: 13299190-13299190 on Build GRCh38
1410417 c.1200G>A p.A400A Chr.20: 13299264-13299264 on Build GRCh38
3939259 c.1328A>T p.Q443L Chr.20: 13299392-13299392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369652412 Chr.20: 13299259-13299259 on Build GRCh38
rs767560781 Chr.20: 13299297-13299297 on Build GRCh38
rs773926392 Chr.20: 13299289-13299289 on Build GRCh38
rs761715304 Chr.20: 13299270-13299270 on Build GRCh38
rs746309693 Chr.20: 13299267-13299267 on Build GRCh38
rs775135191 Chr.20: 13299159-13299159 on Build GRCh38
rs775330290 Chr.20: 13299272-13299272 on Build GRCh38
rs562516676 Chr.20: 13299373-13299373 on Build GRCh38
rs758620056 Chr.20: 13299317-13299317 on Build GRCh38
rs375561259 Chr.20: 13299226-13299226 on Build GRCh38
rs776388531 Chr.20: 13299222-13299222 on Build GRCh38
rs530028092 Chr.20: 13299367-13299367 on Build GRCh38
rs761644256 Chr.20: 13299291-13299291 on Build GRCh38
rs751413226 Chr.20: 13299161-13299161 on Build GRCh38
rs765038981 Chr.20: 13299150-13299150 on Build GRCh38
rs758428662 Chr.20: 13299207-13299207 on Build GRCh38
rs755955610 Chr.20: 13299306-13299306 on Build GRCh38
rs748209062 Chr.20: 13299200-13299200 on Build GRCh38
rs546009979 Chr.20: 13299173-13299173 on Build GRCh38
rs762717119 Chr.20: 13299273-13299273 on Build GRCh38
rs1020887469 Chr.20: 13299371-13299371 on Build GRCh38
rs767160701 Chr.20: 13299170-13299170 on Build GRCh38
rs746194656 Chr.20: 13299366-13299366 on Build GRCh38
rs780571258 Chr.20: 13299372-13299372 on Build GRCh38
rs750157145 Chr.20: 13299300-13299300 on Build GRCh38
rs771478952 Chr.20: 13299246-13299246 on Build GRCh38
rs1010544524 Chr.20: 13299316-13299316 on Build GRCh38
rs541081077 Chr.20: 13299165-13299165 on Build GRCh38
rs757152227 Chr.20: 13299215-13299216 on Build GRCh38
rs781742030 Chr.20: 13299355-13299355 on Build GRCh38
rs757699380 Chr.20: 13299345-13299345 on Build GRCh38
rs763632064 Chr.20: 13299288-13299288 on Build GRCh38
rs747404530 Chr.20: 13299229-13299229 on Build GRCh38
rs779149851 Chr.20: 13299198-13299198 on Build GRCh38
rs200138184 Chr.20: 13299310-13299310 on Build GRCh38
rs755154573 Chr.20: 13299187-13299187 on Build GRCh38
rs752872618 Chr.20: 13299314-13299314 on Build GRCh38

View Full Product Details