Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 180916028-180916281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1614
Entrez Gene ID
57710
Gene Name
KIAA1614
Gene Aliases
-
UniGene
Hs.734816
Gene Chromosome Location
Chr.1: 180913177-180946103 on Build GRCh38
Transcripts
NM_020950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1668248 c.167C>A p.P56H Chr.1: 180916270-180916270 on Build GRCh38
6217222 c.138C>T p.N46N Chr.1: 180916241-180916241 on Build GRCh38
1212254 c.161C>A p.P54H Chr.1: 180916264-180916264 on Build GRCh38
1470155 c.75A>G p.T25T Chr.1: 180916178-180916178 on Build GRCh38
6694355 c.89A>C p.E30A Chr.1: 180916192-180916192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61811249 Chr.1: 180916110-180916110 on Build GRCh38
rs779830320 Chr.1: 180916258-180916258 on Build GRCh38
rs949626029 Chr.1: 180916071-180916071 on Build GRCh38
rs750927784 Chr.1: 180916124-180916124 on Build GRCh38
rs533522709 Chr.1: 180916232-180916232 on Build GRCh38
rs775075372 Chr.1: 180916247-180916247 on Build GRCh38
rs751536042 Chr.1: 180916105-180916105 on Build GRCh38
rs758071065 Chr.1: 180916153-180916153 on Build GRCh38
rs770464783 Chr.1: 180916175-180916175 on Build GRCh38
rs745546581 Chr.1: 180916133-180916136 on Build GRCh38
rs112657356 Chr.1: 180916198-180916198 on Build GRCh38
rs969297428 Chr.1: 180916063-180916063 on Build GRCh38
rs761221405 Chr.1: 180916210-180916210 on Build GRCh38
rs756760078 Chr.1: 180916247-180916247 on Build GRCh38
rs767610080 Chr.1: 180916206-180916206 on Build GRCh38
rs201755087 Chr.1: 180916241-180916241 on Build GRCh38
rs755392688 Chr.1: 180916234-180916234 on Build GRCh38
rs761854753 Chr.1: 180916202-180916202 on Build GRCh38
rs775217655 Chr.1: 180916188-180916188 on Build GRCh38
rs572727647 Chr.1: 180916147-180916147 on Build GRCh38
rs780532411 Chr.1: 180916132-180916132 on Build GRCh38
rs574139354 Chr.1: 180916073-180916073 on Build GRCh38
rs766867679 Chr.1: 180916214-180916214 on Build GRCh38
rs750930526 Chr.1: 180916207-180916207 on Build GRCh38
rs980327415 Chr.1: 180916088-180916088 on Build GRCh38
rs555584886 Chr.1: 180916053-180916053 on Build GRCh38
rs982773933 Chr.1: 180916091-180916091 on Build GRCh38
rs780715348 Chr.1: 180916252-180916252 on Build GRCh38
rs749146657 Chr.1: 180916259-180916259 on Build GRCh38
rs767974924 Chr.1: 180916120-180916120 on Build GRCh38
rs74133666 Chr.1: 180916130-180916130 on Build GRCh38
rs928297408 Chr.1: 180916092-180916092 on Build GRCh38
rs763176337 Chr.1: 180916196-180916196 on Build GRCh38
rs187389379 Chr.1: 180916154-180916154 on Build GRCh38
rs764182204 Chr.1: 180916104-180916104 on Build GRCh38
rs1052135772 Chr.1: 180916185-180916185 on Build GRCh38
rs755988175 Chr.1: 180916253-180916253 on Build GRCh38
rs760035765 Chr.1: 180916262-180916265 on Build GRCh38
rs541265320 Chr.1: 180916102-180916102 on Build GRCh38
rs745788702 Chr.1: 180916184-180916184 on Build GRCh38
rs746570602 Chr.1: 180916174-180916174 on Build GRCh38
rs545594375 Chr.1: 180916096-180916096 on Build GRCh38
rs914145494 Chr.1: 180916070-180916070 on Build GRCh38
rs781042243 Chr.1: 180916177-180916177 on Build GRCh38
rs771572430 Chr.1: 180916246-180916247 on Build GRCh38
rs149264713 Chr.1: 180916187-180916187 on Build GRCh38
rs563705578 Chr.1: 180916176-180916176 on Build GRCh38
rs751204325 Chr.1: 180916245-180916245 on Build GRCh38
rs754193663 Chr.1: 180916227-180916227 on Build GRCh38

View Full Product Details