Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102372008-102372233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18R1
Entrez Gene ID
8809
Gene Name
interleukin 18 receptor 1
Gene Aliases
CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP
UniGene
Hs.469521
Gene Chromosome Location
Chr.2: 102355817-102398777 on Build GRCh38
Transcripts
NM_001282399.1 NM_003855.3 XM_005264039.3 XM_005264040.3 XM_011512099.1 XM_017005181.1 XM_017005182.1 XM_017005183.1 XM_017005184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6509616 c.360C>A p.F120L Chr.2: 102372010-102372010 on Build GRCh38
4914259 c.400A>C p.K134Q Chr.2: 102372050-102372050 on Build GRCh38
4692224 c.404T>G p.F135C Chr.2: 102372054-102372054 on Build GRCh38
5859787 c.416C>T p.T139I Chr.2: 102372066-102372066 on Build GRCh38
1004663 c.401A>C p.K134T Chr.2: 102372051-102372051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904204970 Chr.2: 102372084-102372084 on Build GRCh38
rs375658526 Chr.2: 102372145-102372145 on Build GRCh38
rs929140233 Chr.2: 102372043-102372043 on Build GRCh38
rs754650932 Chr.2: 102372208-102372208 on Build GRCh38
rs758938686 Chr.2: 102372136-102372136 on Build GRCh38
rs781461957 Chr.2: 102372060-102372060 on Build GRCh38
rs770716680 Chr.2: 102372112-102372112 on Build GRCh38
rs766379820 Chr.2: 102372040-102372040 on Build GRCh38
rs776004141 Chr.2: 102372114-102372114 on Build GRCh38
rs545609877 Chr.2: 102372178-102372178 on Build GRCh38
rs754016574 Chr.2: 102372052-102372052 on Build GRCh38
rs1008028202 Chr.2: 102372085-102372085 on Build GRCh38
rs34216045 Chr.2: 102372066-102372066 on Build GRCh38
rs890041500 Chr.2: 102372201-102372201 on Build GRCh38
rs901806108 Chr.2: 102372169-102372169 on Build GRCh38
rs758228602 Chr.2: 102372095-102372095 on Build GRCh38
rs745445571 Chr.2: 102372136-102372136 on Build GRCh38
rs746601953 Chr.2: 102372098-102372098 on Build GRCh38
rs778956300 Chr.2: 102372065-102372065 on Build GRCh38
rs774878937 Chr.2: 102372153-102372153 on Build GRCh38

View Full Product Details