Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39357174-39357435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
UniGene
Hs.588202
Gene Chromosome Location
Chr.6: 39335100-39725405 on Build GRCh38
Transcripts
NM_001289020.1 NM_001289021.1 NM_001289024.1 NM_145027.4 XM_005248904.4 XM_011514357.2 XM_011514358.2 XM_011514359.2 XM_017010427.1 XM_017010428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3830458 c.1941C>T p.L647L Chr.6: 39357348-39357348 on Build GRCh38
3626985 c.2152G>A p.E718K Chr.6: 39357305-39357305 on Build GRCh38
5771703 c.2094A>T p.P698P Chr.6: 39357363-39357363 on Build GRCh38
7181129 c.483C>G p.L161L Chr.6: 39357327-39357327 on Build GRCh38
3830459 c.462C>T p.L154L Chr.6: 39357348-39357348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757043014 Chr.6: 39357233-39357233 on Build GRCh38
rs761988781 Chr.6: 39357375-39357375 on Build GRCh38
rs771315080 Chr.6: 39357259-39357259 on Build GRCh38
rs149250289 Chr.6: 39357296-39357296 on Build GRCh38
rs543067276 Chr.6: 39357301-39357301 on Build GRCh38
rs769555494 Chr.6: 39357238-39357238 on Build GRCh38
rs779040500 Chr.6: 39357381-39357381 on Build GRCh38
rs753922680 Chr.6: 39357388-39357389 on Build GRCh38
rs920255452 Chr.6: 39357287-39357287 on Build GRCh38
rs768278517 Chr.6: 39357322-39357322 on Build GRCh38
rs571705140 Chr.6: 39357201-39357201 on Build GRCh38
rs78761242 Chr.6: 39357258-39357258 on Build GRCh38
rs774117259 Chr.6: 39357257-39357257 on Build GRCh38
rs776804143 Chr.6: 39357281-39357281 on Build GRCh38
rs199509937 Chr.6: 39357309-39357309 on Build GRCh38
rs868017412 Chr.6: 39357315-39357315 on Build GRCh38
rs996091448 Chr.6: 39357209-39357209 on Build GRCh38
rs748762128 Chr.6: 39357248-39357248 on Build GRCh38
rs958226159 Chr.6: 39357208-39357208 on Build GRCh38
rs201592417 Chr.6: 39357340-39357340 on Build GRCh38
rs930456055 Chr.6: 39357326-39357326 on Build GRCh38
rs767589890 Chr.6: 39357400-39357400 on Build GRCh38
rs765692235 Chr.6: 39357291-39357291 on Build GRCh38
rs772948774 Chr.6: 39357339-39357339 on Build GRCh38
rs763515741 Chr.6: 39357348-39357348 on Build GRCh38
rs764196404 Chr.6: 39357300-39357300 on Build GRCh38
rs781485675 Chr.6: 39357306-39357306 on Build GRCh38
rs375678835 Chr.6: 39357365-39357365 on Build GRCh38
rs967371660 Chr.6: 39357248-39357249 on Build GRCh38
rs778625487 Chr.6: 39357331-39357331 on Build GRCh38
rs935419009 Chr.6: 39357372-39357372 on Build GRCh38
rs371202903 Chr.6: 39357236-39357236 on Build GRCh38
rs768333480 Chr.6: 39357252-39357252 on Build GRCh38
rs780176724 Chr.6: 39357244-39357244 on Build GRCh38
rs749280102 Chr.6: 39357319-39357319 on Build GRCh38
rs759949178 Chr.6: 39357282-39357282 on Build GRCh38
rs144915881 Chr.6: 39357226-39357226 on Build GRCh38
rs751825840 Chr.6: 39357232-39357232 on Build GRCh38
rs372001923 Chr.6: 39357335-39357335 on Build GRCh38
rs20455 Chr.6: 39357302-39357302 on Build GRCh38
rs774232734 Chr.6: 39357362-39357362 on Build GRCh38
rs777920571 Chr.6: 39357230-39357230 on Build GRCh38
rs764141442 Chr.6: 39357349-39357349 on Build GRCh38
rs893806515 Chr.6: 39357402-39357402 on Build GRCh38
rs775941976 Chr.6: 39357346-39357346 on Build GRCh38
rs902577945 Chr.6: 39357360-39357360 on Build GRCh38
rs960435080 Chr.6: 39357294-39357294 on Build GRCh38

View Full Product Details