Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 66922499-66922758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPHN
Entrez Gene ID
10243
Gene Name
gephyrin
Gene Aliases
GEPH, GPH, GPHRYN, HKPX1, MOCODC
UniGene
Hs.208765
Gene Chromosome Location
Chr.14: 66507407-67735831 on Build GRCh38
Transcripts
NM_001024218.1 NM_020806.4 XM_005267254.3 XM_011536340.2 XM_011536342.2 XM_011536343.2 XM_011536344.2 XM_011536345.2 XM_011536346.2 XM_011536347.2 XM_017020913.1 XM_017020914.1 XM_017020915.1 XM_017020916.1 XM_017020917.1 XM_017020918.1 XM_017020919.1 XM_017020920.1 XM_017020921.1 XM_017020922.1 XM_017020923.1 XM_017020924.1 XM_017020925.1 XM_017020926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4913386 c.544G>T p.E182* Chr.14: 66922753-66922753 on Build GRCh38
5382531 c.502C>T p.L168F Chr.14: 66922711-66922711 on Build GRCh38
4051919 c.515C>T p.A172V Chr.14: 66922724-66922724 on Build GRCh38
3497223 c.466C>T p.Q156* Chr.14: 66922675-66922675 on Build GRCh38
3419969 c.499G>T p.D167Y Chr.14: 66922708-66922708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749859135 Chr.14: 66922710-66922710 on Build GRCh38
rs928365129 Chr.14: 66922559-66922559 on Build GRCh38
rs910287556 Chr.14: 66922569-66922569 on Build GRCh38
rs775343201 Chr.14: 66922574-66922574 on Build GRCh38
rs761317030 Chr.14: 66922667-66922667 on Build GRCh38
rs558954236 Chr.14: 66922538-66922538 on Build GRCh38
rs768057286 Chr.14: 66922717-66922717 on Build GRCh38
rs889646027 Chr.14: 66922606-66922606 on Build GRCh38
rs765769429 Chr.14: 66922639-66922640 on Build GRCh38
rs74936384 Chr.14: 66922629-66922629 on Build GRCh38
rs552614595 Chr.14: 66922587-66922587 on Build GRCh38
rs551889185 Chr.14: 66922584-66922584 on Build GRCh38
rs768570275 Chr.14: 66922660-66922660 on Build GRCh38
rs771045451 Chr.14: 66922624-66922624 on Build GRCh38
rs78369678 Chr.14: 66922586-66922586 on Build GRCh38
rs776431468 Chr.14: 66922663-66922663 on Build GRCh38
rs112443999 Chr.14: 66922557-66922557 on Build GRCh38
rs764823306 Chr.14: 66922705-66922705 on Build GRCh38
rs745850208 Chr.14: 66922637-66922637 on Build GRCh38
rs527242930 Chr.14: 66922571-66922571 on Build GRCh38
rs930238508 Chr.14: 66922601-66922601 on Build GRCh38
rs966531598 Chr.14: 66922651-66922651 on Build GRCh38
rs541706472 Chr.14: 66922545-66922545 on Build GRCh38
rs956810579 Chr.14: 66922530-66922530 on Build GRCh38
rs373140442 Chr.14: 66922592-66922592 on Build GRCh38
rs371904779 Chr.14: 66922718-66922718 on Build GRCh38
rs770031914 Chr.14: 66922541-66922541 on Build GRCh38
rs377162663 Chr.14: 66922640-66922640 on Build GRCh38
rs773936647 Chr.14: 66922640-66922640 on Build GRCh38
rs775546697 Chr.14: 66922649-66922649 on Build GRCh38
rs760605677 Chr.14: 66922653-66922653 on Build GRCh38
rs948845291 Chr.14: 66922542-66922542 on Build GRCh38
rs771946114 Chr.14: 66922639-66922639 on Build GRCh38
rs990369556 Chr.14: 66922529-66922529 on Build GRCh38

View Full Product Details