Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 188570069-188570312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GULP1
Entrez Gene ID
51454
Gene Name
GULP, engulfment adaptor PTB domain containing 1
Gene Aliases
CED-6, CED6, GULP
UniGene
Hs.470887
Gene Chromosome Location
Chr.2: 188291669-188595926 on Build GRCh38
Transcripts
NM_001252668.1 NM_001252669.1 NM_016315.3 NR_045562.1 NR_045563.1 XM_006712580.3 XM_006712581.1 XM_006712582.1 XM_006712583.3 XM_006712584.3 XM_006712585.1 XM_006712589.3 XM_011511329.1 XM_011511331.1 XM_011511332.1 XM_011511333.1 XM_011511335.2 XM_017004303.1 XM_017004304.1 XM_017004305.1 XM_017004306.1 XM_017004307.1 XM_017004308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1614044 c.593A>C p.Q198P Chr.2: 188570104-188570104 on Build GRCh38
1013927 c.561A>G p.V187V Chr.2: 188570072-188570072 on Build GRCh38
1403984 c.580C>T p.L194L Chr.2: 188570091-188570091 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775067927 Chr.2: 188570152-188570152 on Build GRCh38
rs911500013 Chr.2: 188570263-188570263 on Build GRCh38
rs768387068 Chr.2: 188570133-188570133 on Build GRCh38
rs777750297 Chr.2: 188570174-188570174 on Build GRCh38
rs200711089 Chr.2: 188570157-188570157 on Build GRCh38
rs904193397 Chr.2: 188570165-188570165 on Build GRCh38
rs757385151 Chr.2: 188570172-188570172 on Build GRCh38
rs996369599 Chr.2: 188570213-188570213 on Build GRCh38
rs187302982 Chr.2: 188570156-188570156 on Build GRCh38
rs746688944 Chr.2: 188570128-188570128 on Build GRCh38
rs778832408 Chr.2: 188570116-188570116 on Build GRCh38
rs115951524 Chr.2: 188570276-188570276 on Build GRCh38
rs746931879 Chr.2: 188570184-188570184 on Build GRCh38
rs183573379 Chr.2: 188570264-188570264 on Build GRCh38
rs373024772 Chr.2: 188570149-188570149 on Build GRCh38
rs968823559 Chr.2: 188570271-188570271 on Build GRCh38
rs977297425 Chr.2: 188570266-188570266 on Build GRCh38
rs191741660 Chr.2: 188570259-188570259 on Build GRCh38
rs771919175 Chr.2: 188570119-188570119 on Build GRCh38
rs1037869280 Chr.2: 188570160-188570160 on Build GRCh38
rs1046868113 Chr.2: 188570138-188570138 on Build GRCh38
rs770441140 Chr.2: 188570236-188570236 on Build GRCh38
rs547757588 Chr.2: 188570225-188570225 on Build GRCh38
rs748053743 Chr.2: 188570231-188570231 on Build GRCh38
rs756390336 Chr.2: 188570169-188570169 on Build GRCh38
rs771729850 Chr.2: 188570150-188570150 on Build GRCh38
rs374345703 Chr.2: 188570170-188570170 on Build GRCh38
rs994268984 Chr.2: 188570254-188570254 on Build GRCh38
rs985666525 Chr.2: 188570260-188570260 on Build GRCh38
rs776083655 Chr.2: 188570147-188570147 on Build GRCh38
rs912951508 Chr.2: 188570111-188570111 on Build GRCh38
rs1014527621 Chr.2: 188570212-188570212 on Build GRCh38
rs1026901841 Chr.2: 188570258-188570258 on Build GRCh38
rs775234108 Chr.2: 188570122-188570122 on Build GRCh38
rs753123839 Chr.2: 188570166-188570166 on Build GRCh38
rs143831338 Chr.2: 188570106-188570106 on Build GRCh38

View Full Product Details