Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92811426-92811651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA5
Entrez Gene ID
9950
Gene Name
golgin A5
Gene Aliases
GOLIM5, RFG5, ret-II
UniGene
Hs.104320
Gene Chromosome Location
Chr.14: 92794231-92839963 on Build GRCh38
Transcripts
NM_005113.3 XM_011537420.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1735607 c.1156C>T p.R386C Chr.14: 92811590-92811590 on Build GRCh38
6328165 c.1201T>A p.S401T Chr.14: 92811635-92811635 on Build GRCh38
1167573 c.1208_1210delAGA p.K405delK Chr.14: 92811642-92811644 on Build GRCh38
2252111 c.1154A>G p.E385G Chr.14: 92811588-92811588 on Build GRCh38
6326808 c.1168G>A p.A390T Chr.14: 92811602-92811602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963552783 Chr.14: 92811509-92811509 on Build GRCh38
rs994209838 Chr.14: 92811491-92811491 on Build GRCh38
rs536029459 Chr.14: 92811471-92811471 on Build GRCh38
rs768382436 Chr.14: 92811567-92811567 on Build GRCh38
rs371221028 Chr.14: 92811553-92811553 on Build GRCh38
rs375012765 Chr.14: 92811520-92811520 on Build GRCh38
rs35865539 Chr.14: 92811573-92811573 on Build GRCh38
rs1025195467 Chr.14: 92811474-92811474 on Build GRCh38
rs988707760 Chr.14: 92811587-92811587 on Build GRCh38
rs766924395 Chr.14: 92811611-92811611 on Build GRCh38
rs564154198 Chr.14: 92811527-92811527 on Build GRCh38
rs780935662 Chr.14: 92811548-92811548 on Build GRCh38
rs189037186 Chr.14: 92811480-92811480 on Build GRCh38
rs755174731 Chr.14: 92811526-92811526 on Build GRCh38
rs543664165 Chr.14: 92811617-92811617 on Build GRCh38
rs553333516 Chr.14: 92811563-92811563 on Build GRCh38
rs772660824 Chr.14: 92811590-92811590 on Build GRCh38
rs200288795 Chr.14: 92811591-92811591 on Build GRCh38
rs981549401 Chr.14: 92811531-92811531 on Build GRCh38
rs370258336 Chr.14: 92811516-92811516 on Build GRCh38
rs112256540 Chr.14: 92811547-92811547 on Build GRCh38
rs771613439 Chr.14: 92811588-92811588 on Build GRCh38
rs531515677 Chr.14: 92811544-92811544 on Build GRCh38
rs371151849 Chr.14: 92811612-92811612 on Build GRCh38
rs866064181 Chr.14: 92811615-92811615 on Build GRCh38
rs984132946 Chr.14: 92811551-92811551 on Build GRCh38
rs759928094 Chr.14: 92811614-92811614 on Build GRCh38
rs990953321 Chr.14: 92811512-92811512 on Build GRCh38
rs142273431 Chr.14: 92811554-92811554 on Build GRCh38
rs751943504 Chr.14: 92811566-92811566 on Build GRCh38
rs760878925 Chr.14: 92811514-92811514 on Build GRCh38
rs774167940 Chr.14: 92811571-92811571 on Build GRCh38
rs752736005 Chr.14: 92811535-92811535 on Build GRCh38
rs192692726 Chr.14: 92811533-92811533 on Build GRCh38
rs368617884 Chr.14: 92811503-92811503 on Build GRCh38
rs746430230 Chr.14: 92811522-92811522 on Build GRCh38

View Full Product Details