Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 67338539-67338796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12RB2
Entrez Gene ID
3595
Gene Name
interleukin 12 receptor subunit beta 2
Gene Aliases
-
UniGene
Hs.479347
Gene Chromosome Location
Chr.1: 67307351-67397730 on Build GRCh38
Transcripts
NM_001258214.1 NM_001258215.1 NM_001258216.1 NM_001319233.1 NM_001559.2 NR_047583.1 NR_047584.1 XM_005270825.2 XM_005270827.2 XM_005270828.3 XM_006710617.2 XM_011541383.2 XM_011541384.2 XM_017001202.1 XM_017001203.1 XM_017001204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
682321 c.961C>T p.P321S Chr.1: 67338626-67338626 on Build GRCh38
911834 c.962C>A p.P321H Chr.1: 67338627-67338627 on Build GRCh38
6684863 c.994C>T p.R332W Chr.1: 67338659-67338659 on Build GRCh38
6063872 c.1038G>A p.K346K Chr.1: 67338703-67338703 on Build GRCh38
6540523 c.979G>T p.V327F Chr.1: 67338644-67338644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771137126 Chr.1: 67338652-67338652 on Build GRCh38
rs990421118 Chr.1: 67338705-67338705 on Build GRCh38
rs556830305 Chr.1: 67338613-67338613 on Build GRCh38
rs749883157 Chr.1: 67338688-67338688 on Build GRCh38
rs201336326 Chr.1: 67338666-67338666 on Build GRCh38
rs756830105 Chr.1: 67338574-67338574 on Build GRCh38
rs559654679 Chr.1: 67338610-67338610 on Build GRCh38
rs755553813 Chr.1: 67338692-67338692 on Build GRCh38
rs150877640 Chr.1: 67338653-67338653 on Build GRCh38
rs747266023 Chr.1: 67338649-67338649 on Build GRCh38
rs943199534 Chr.1: 67338658-67338658 on Build GRCh38
rs772946364 Chr.1: 67338597-67338598 on Build GRCh38
rs779361557 Chr.1: 67338736-67338736 on Build GRCh38
rs750094276 Chr.1: 67338589-67338589 on Build GRCh38
rs1014580443 Chr.1: 67338642-67338642 on Build GRCh38
rs779538669 Chr.1: 67338627-67338627 on Build GRCh38
rs748707538 Chr.1: 67338659-67338659 on Build GRCh38
rs767005706 Chr.1: 67338579-67338579 on Build GRCh38
rs529800718 Chr.1: 67338764-67338764 on Build GRCh38
rs748668436 Chr.1: 67338744-67338744 on Build GRCh38
rs748649979 Chr.1: 67338629-67338629 on Build GRCh38
rs761684036 Chr.1: 67338668-67338668 on Build GRCh38
rs762742625 Chr.1: 67338626-67338626 on Build GRCh38
rs887453058 Chr.1: 67338636-67338636 on Build GRCh38
rs148793796 Chr.1: 67338634-67338634 on Build GRCh38
rs780301931 Chr.1: 67338591-67338591 on Build GRCh38
rs578200723 Chr.1: 67338637-67338638 on Build GRCh38
rs190952170 Chr.1: 67338612-67338612 on Build GRCh38
rs751448905 Chr.1: 67338708-67338709 on Build GRCh38
rs528852703 Chr.1: 67338762-67338762 on Build GRCh38
rs778022576 Chr.1: 67338761-67338761 on Build GRCh38
rs763794469 Chr.1: 67338660-67338660 on Build GRCh38
rs771579027 Chr.1: 67338628-67338628 on Build GRCh38
rs758934202 Chr.1: 67338753-67338753 on Build GRCh38
rs755747280 Chr.1: 67338590-67338590 on Build GRCh38
rs753425797 Chr.1: 67338716-67338716 on Build GRCh38
rs772290732 Chr.1: 67338633-67338633 on Build GRCh38
rs755427561 Chr.1: 67338717-67338717 on Build GRCh38
rs781546820 Chr.1: 67338768-67338768 on Build GRCh38
rs866487660 Chr.1: 67338690-67338690 on Build GRCh38
rs146568326 Chr.1: 67338672-67338672 on Build GRCh38

View Full Product Details