Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102015752-102015983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1R2
Entrez Gene ID
7850
Gene Name
interleukin 1 receptor type 2
Gene Aliases
CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB
UniGene
Hs.25333
Gene Chromosome Location
Chr.2: 101991805-102028544 on Build GRCh38
Transcripts
NM_001261419.1 NM_004633.3 NR_048564.1 XM_006712734.3 XM_006712736.3 XM_011511801.2 XM_011511803.2 XM_011511804.2 XM_011511805.2 XM_011511807.1 XM_017004889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1210544 c.445G>T p.V149L Chr.2: 102015983-102015983 on Build GRCh38
5025682 c.369C>T p.L123L Chr.2: 102015907-102015907 on Build GRCh38
4759576 c.414C>A p.Y138* Chr.2: 102015952-102015952 on Build GRCh38
3726373 c.383A>G p.N128S Chr.2: 102015921-102015921 on Build GRCh38
4759575 c.409T>C p.S137P Chr.2: 102015947-102015947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190112469 Chr.2: 102015895-102015895 on Build GRCh38
rs770663605 Chr.2: 102015905-102015905 on Build GRCh38
rs757210661 Chr.2: 102015906-102015906 on Build GRCh38
rs766619759 Chr.2: 102015840-102015840 on Build GRCh38
rs746839384 Chr.2: 102015897-102015897 on Build GRCh38
rs766781084 Chr.2: 102015957-102015957 on Build GRCh38
rs548206510 Chr.2: 102015885-102015885 on Build GRCh38
rs754101980 Chr.2: 102015849-102015849 on Build GRCh38
rs536898115 Chr.2: 102015940-102015940 on Build GRCh38
rs768129599 Chr.2: 102015880-102015880 on Build GRCh38
rs758364548 Chr.2: 102015866-102015866 on Build GRCh38
rs150767253 Chr.2: 102015952-102015952 on Build GRCh38
rs780957914 Chr.2: 102015921-102015921 on Build GRCh38
rs762159296 Chr.2: 102015947-102015947 on Build GRCh38
rs752575255 Chr.2: 102015862-102015862 on Build GRCh38
rs751074815 Chr.2: 102015848-102015849 on Build GRCh38
rs759979263 Chr.2: 102015875-102015882 on Build GRCh38
rs779024862 Chr.2: 102015861-102015861 on Build GRCh38
rs760997000 Chr.2: 102015955-102015955 on Build GRCh38
rs148689791 Chr.2: 102015939-102015939 on Build GRCh38
rs113418071 Chr.2: 102015810-102015810 on Build GRCh38
rs1056231350 Chr.2: 102015928-102015928 on Build GRCh38
rs144052963 Chr.2: 102015954-102015954 on Build GRCh38
rs1031997209 Chr.2: 102015903-102015903 on Build GRCh38
rs563841433 Chr.2: 102015849-102015849 on Build GRCh38
rs755029686 Chr.2: 102015859-102015859 on Build GRCh38
rs745562227 Chr.2: 102015935-102015935 on Build GRCh38
rs571407860 Chr.2: 102015849-102015850 on Build GRCh38

View Full Product Details