Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167054895-167055168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPA33
Entrez Gene ID
10223
Gene Name
glycoprotein A33
Gene Aliases
A33
UniGene
Hs.651244
Gene Chromosome Location
Chr.1: 167052836-167090631 on Build GRCh38
Transcripts
NM_005814.2 XM_017000005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4687705 c.795C>T p.D265D Chr.1: 167055008-167055008 on Build GRCh38
4559028 c.789G>A p.G263G Chr.1: 167055014-167055014 on Build GRCh38
899014 c.724G>A p.A242T Chr.1: 167055079-167055079 on Build GRCh38
6122380 c.827+1G>T p.? Chr.1: 167054975-167054975 on Build GRCh38
899013 c.767T>C p.I256T Chr.1: 167055036-167055036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747676758 Chr.1: 167055100-167055100 on Build GRCh38
rs539638930 Chr.1: 167054946-167054946 on Build GRCh38
rs762795168 Chr.1: 167055037-167055037 on Build GRCh38
rs889351816 Chr.1: 167054926-167054926 on Build GRCh38
rs964109007 Chr.1: 167055106-167055106 on Build GRCh38
rs775149743 Chr.1: 167055140-167055140 on Build GRCh38
rs750965272 Chr.1: 167055077-167055077 on Build GRCh38
rs757840288 Chr.1: 167055072-167055072 on Build GRCh38
rs138343655 Chr.1: 167055079-167055079 on Build GRCh38
rs181989930 Chr.1: 167054944-167054944 on Build GRCh38
rs201186970 Chr.1: 167055078-167055078 on Build GRCh38
rs749148901 Chr.1: 167055124-167055124 on Build GRCh38
rs755457133 Chr.1: 167054976-167054976 on Build GRCh38
rs773966640 Chr.1: 167055128-167055128 on Build GRCh38
rs758558541 Chr.1: 167054989-167054989 on Build GRCh38
rs562657939 Chr.1: 167055127-167055127 on Build GRCh38
rs773460511 Chr.1: 167055005-167055007 on Build GRCh38
rs755117935 Chr.1: 167055097-167055097 on Build GRCh38
rs373464180 Chr.1: 167054961-167054961 on Build GRCh38
rs748117218 Chr.1: 167055018-167055018 on Build GRCh38
rs773200729 Chr.1: 167055114-167055114 on Build GRCh38
rs759260885 Chr.1: 167055130-167055130 on Build GRCh38
rs989577393 Chr.1: 167055085-167055085 on Build GRCh38
rs769850908 Chr.1: 167055105-167055105 on Build GRCh38
rs765041543 Chr.1: 167054934-167054934 on Build GRCh38
rs747279502 Chr.1: 167055081-167055081 on Build GRCh38
rs759087048 Chr.1: 167055068-167055068 on Build GRCh38
rs771554012 Chr.1: 167055132-167055132 on Build GRCh38
rs377334435 Chr.1: 167054943-167054943 on Build GRCh38
rs761718786 Chr.1: 167054927-167054927 on Build GRCh38
rs776526472 Chr.1: 167055016-167055016 on Build GRCh38
rs146541901 Chr.1: 167055055-167055055 on Build GRCh38
rs752212756 Chr.1: 167055071-167055071 on Build GRCh38
rs763671076 Chr.1: 167055142-167055142 on Build GRCh38
rs371212925 Chr.1: 167054949-167054949 on Build GRCh38
rs150875349 Chr.1: 167054975-167054975 on Build GRCh38
rs1003739926 Chr.1: 167055064-167055064 on Build GRCh38
rs201698271 Chr.1: 167055052-167055052 on Build GRCh38
rs767074475 Chr.1: 167055070-167055070 on Build GRCh38
rs200952830 Chr.1: 167055101-167055101 on Build GRCh38
rs148037017 Chr.1: 167055007-167055007 on Build GRCh38
rs368155919 Chr.1: 167055008-167055008 on Build GRCh38
rs773004684 Chr.1: 167055019-167055019 on Build GRCh38
rs866381875 Chr.1: 167055050-167055050 on Build GRCh38
rs199580563 Chr.1: 167055149-167055149 on Build GRCh38
rs370661795 Chr.1: 167055010-167055010 on Build GRCh38
rs148908812 Chr.1: 167055005-167055005 on Build GRCh38
rs765517721 Chr.1: 167054973-167054974 on Build GRCh38
rs748168572 Chr.1: 167055103-167055103 on Build GRCh38
rs766905165 Chr.1: 167054952-167054952 on Build GRCh38
rs765778378 Chr.1: 167055076-167055076 on Build GRCh38
rs375793730 Chr.1: 167055126-167055126 on Build GRCh38
rs751157899 Chr.1: 167054947-167054947 on Build GRCh38
rs368510814 Chr.1: 167055123-167055123 on Build GRCh38
rs369432393 Chr.1: 167055141-167055141 on Build GRCh38

View Full Product Details