Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87372638-87372889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B11
Entrez Gene ID
51170
Gene Name
hydroxysteroid 17-beta dehydrogenase 11
Gene Aliases
17-BETA-HSD11, 17-BETA-HSDXI, 17BHSD11, DHRS8, PAN1B, RETSDR2, SDR16C2
UniGene
Hs.594923
Gene Chromosome Location
Chr.4: 87336522-87391303 on Build GRCh38
Transcripts
NM_016245.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792749 c.487A>T p.N163Y Chr.4: 87372779-87372779 on Build GRCh38
4126571 c.476T>C p.M159T Chr.4: 87372790-87372790 on Build GRCh38
481656 c.470C>T p.P157L Chr.4: 87372796-87372796 on Build GRCh38
5866905 c.511G>A p.A171T Chr.4: 87372755-87372755 on Build GRCh38
6682334 c.454A>G p.T152A Chr.4: 87372812-87372812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763299916 Chr.4: 87372721-87372723 on Build GRCh38
rs771233784 Chr.4: 87372760-87372761 on Build GRCh38
rs746954003 Chr.4: 87372762-87372762 on Build GRCh38
rs751993837 Chr.4: 87372697-87372697 on Build GRCh38
rs771878491 Chr.4: 87372748-87372748 on Build GRCh38
rs1031742679 Chr.4: 87372751-87372751 on Build GRCh38
rs6531985 Chr.4: 87372735-87372735 on Build GRCh38
rs774254146 Chr.4: 87372763-87372764 on Build GRCh38
rs748027042 Chr.4: 87372739-87372739 on Build GRCh38
rs765907071 Chr.4: 87372681-87372681 on Build GRCh38
rs397724448 Chr.4: 87372846-87372847 on Build GRCh38
rs755298883 Chr.4: 87372733-87372733 on Build GRCh38
rs770798852 Chr.4: 87372772-87372772 on Build GRCh38
rs139110766 Chr.4: 87372786-87372786 on Build GRCh38
rs200974821 Chr.4: 87372774-87372774 on Build GRCh38
rs376839018 Chr.4: 87372689-87372689 on Build GRCh38
rs775026637 Chr.4: 87372789-87372789 on Build GRCh38
rs11375790 Chr.4: 87372840-87372841 on Build GRCh38
rs767942962 Chr.4: 87372719-87372719 on Build GRCh38
rs767996059 Chr.4: 87372829-87372829 on Build GRCh38
rs753994183 Chr.4: 87372850-87372850 on Build GRCh38
rs540105188 Chr.4: 87372683-87372683 on Build GRCh38
rs372213288 Chr.4: 87372729-87372729 on Build GRCh38
rs757538154 Chr.4: 87372716-87372716 on Build GRCh38
rs114331860 Chr.4: 87372732-87372732 on Build GRCh38
rs759240314 Chr.4: 87372686-87372686 on Build GRCh38
rs761317660 Chr.4: 87372845-87372845 on Build GRCh38
rs376344070 Chr.4: 87372849-87372849 on Build GRCh38
rs750756975 Chr.4: 87372842-87372842 on Build GRCh38
rs765150541 Chr.4: 87372750-87372750 on Build GRCh38
rs1029887284 Chr.4: 87372799-87372799 on Build GRCh38
rs750995514 Chr.4: 87372720-87372720 on Build GRCh38
rs766984195 Chr.4: 87372847-87372847 on Build GRCh38
rs762653576 Chr.4: 87372825-87372825 on Build GRCh38
rs769534010 Chr.4: 87372787-87372787 on Build GRCh38
rs760559491 Chr.4: 87372668-87372668 on Build GRCh38
rs895669004 Chr.4: 87372667-87372667 on Build GRCh38
rs896955005 Chr.4: 87372705-87372705 on Build GRCh38
rs201766591 Chr.4: 87372691-87372691 on Build GRCh38

View Full Product Details