Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64293048-64293307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK4
Entrez Gene ID
50801
Gene Name
potassium two pore domain channel subfamily K member 4
Gene Aliases
K2p4.1, TRAAK, TRAAK1
UniGene
Hs.647233
Gene Chromosome Location
Chr.11: 64291302-64300031 on Build GRCh38
Transcripts
NM_001317090.1 NM_033310.2 NR_133660.1 NR_133661.1 NR_133662.1

Gene Symbol
KCNK4-TEX40
Entrez Gene ID
106780802
Gene Name
KCNK4-TEX40 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 64291722-64304769 on Build GRCh38
Transcripts
NM_001317090.1 NM_033310.2 NR_133660.1 NR_133661.1 NR_133662.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4468172 c.152C>T p.P51L Chr.11: 64293170-64293170 on Build GRCh38
4458063 c.106C>T p.Q36* Chr.11: 64293124-64293124 on Build GRCh38
6413354 c.163G>A p.D55N Chr.11: 64293181-64293181 on Build GRCh38
6383786 c.58G>A p.G20S Chr.11: 64293076-64293076 on Build GRCh38
5473188 c.36G>A p.L12L Chr.11: 64293054-64293054 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757639151 Chr.11: 64293171-64293171 on Build GRCh38
rs749434320 Chr.11: 64293225-64293225 on Build GRCh38
rs1000079532 Chr.11: 64293184-64293184 on Build GRCh38
rs759496072 Chr.11: 64293094-64293094 on Build GRCh38
rs150371580 Chr.11: 64293146-64293146 on Build GRCh38
rs989589691 Chr.11: 64293222-64293222 on Build GRCh38
rs993140819 Chr.11: 64293082-64293082 on Build GRCh38
rs574534061 Chr.11: 64293194-64293194 on Build GRCh38
rs763517876 Chr.11: 64293132-64293132 on Build GRCh38
rs35930898 Chr.11: 64293227-64293227 on Build GRCh38
rs962540686 Chr.11: 64293092-64293092 on Build GRCh38
rs80251760 Chr.11: 64293245-64293245 on Build GRCh38
rs754420092 Chr.11: 64293202-64293202 on Build GRCh38
rs57301737 Chr.11: 64293170-64293170 on Build GRCh38
rs1017472172 Chr.11: 64293158-64293158 on Build GRCh38
rs1046196335 Chr.11: 64293251-64293251 on Build GRCh38
rs746082000 Chr.11: 64293078-64293078 on Build GRCh38
rs779639783 Chr.11: 64293181-64293181 on Build GRCh38
rs753131235 Chr.11: 64293123-64293123 on Build GRCh38
rs986784603 Chr.11: 64293167-64293167 on Build GRCh38
rs954942404 Chr.11: 64293156-64293156 on Build GRCh38
rs958013446 Chr.11: 64293210-64293210 on Build GRCh38
rs576890358 Chr.11: 64293281-64293281 on Build GRCh38
rs913967603 Chr.11: 64293242-64293242 on Build GRCh38
rs778547469 Chr.11: 64293223-64293223 on Build GRCh38
rs1015277642 Chr.11: 64293081-64293081 on Build GRCh38
rs201624439 Chr.11: 64293144-64293144 on Build GRCh38
rs563276649 Chr.11: 64293257-64293257 on Build GRCh38
rs202162247 Chr.11: 64293111-64293111 on Build GRCh38
rs1037695985 Chr.11: 64293072-64293072 on Build GRCh38

View Full Product Details