Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67435896-67436135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD11B2
Entrez Gene ID
3291
Gene Name
hydroxysteroid 11-beta dehydrogenase 2
Gene Aliases
AME, AME1, HSD11K, HSD2, SDR9C3
UniGene
Hs.1376
Gene Chromosome Location
Chr.16: 67431133-67437553 on Build GRCh38
Transcripts
NM_000196.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972443 c.518T>C p.V173A Chr.16: 67435996-67435996 on Build GRCh38
4691255 c.643G>A p.V215M Chr.16: 67436121-67436121 on Build GRCh38
6682323 c.592G>A p.E198K Chr.16: 67436070-67436070 on Build GRCh38
1378928 c.586G>A p.A196T Chr.16: 67436064-67436064 on Build GRCh38
4061854 c.616C>T p.L206L Chr.16: 67436094-67436094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368904749 Chr.16: 67435931-67435931 on Build GRCh38
rs778006489 Chr.16: 67435996-67435996 on Build GRCh38
rs142435782 Chr.16: 67435971-67435971 on Build GRCh38
rs747711207 Chr.16: 67436044-67436044 on Build GRCh38
rs138853656 Chr.16: 67435990-67435990 on Build GRCh38
rs5480 Chr.16: 67436066-67436066 on Build GRCh38
rs767302787 Chr.16: 67435943-67435943 on Build GRCh38
rs45483293 Chr.16: 67436012-67436012 on Build GRCh38
rs149312221 Chr.16: 67436075-67436075 on Build GRCh38
rs72650120 Chr.16: 67435945-67435945 on Build GRCh38
rs747223797 Chr.16: 67435998-67435998 on Build GRCh38
rs994079941 Chr.16: 67436090-67436091 on Build GRCh38
rs765709883 Chr.16: 67436065-67436065 on Build GRCh38
rs765711669 Chr.16: 67435991-67435991 on Build GRCh38
rs752793148 Chr.16: 67435994-67435994 on Build GRCh38
rs560632514 Chr.16: 67436064-67436064 on Build GRCh38
rs544892891 Chr.16: 67435984-67435984 on Build GRCh38
rs369707935 Chr.16: 67435979-67435979 on Build GRCh38
rs375912667 Chr.16: 67435915-67435915 on Build GRCh38
rs761667105 Chr.16: 67436007-67436007 on Build GRCh38
rs868377154 Chr.16: 67436036-67436036 on Build GRCh38
rs563098276 Chr.16: 67435995-67435995 on Build GRCh38
rs918770228 Chr.16: 67436035-67436035 on Build GRCh38
rs199824469 Chr.16: 67435970-67435970 on Build GRCh38
rs542287681 Chr.16: 67436063-67436063 on Build GRCh38
rs889533349 Chr.16: 67436021-67436021 on Build GRCh38
rs749168334 Chr.16: 67435920-67435920 on Build GRCh38
rs773066458 Chr.16: 67436060-67436060 on Build GRCh38
rs774383256 Chr.16: 67436040-67436040 on Build GRCh38
rs779034293 Chr.16: 67436116-67436116 on Build GRCh38
rs768507002 Chr.16: 67436034-67436034 on Build GRCh38
rs142073122 Chr.16: 67436009-67436009 on Build GRCh38
rs764141451 Chr.16: 67436070-67436070 on Build GRCh38
rs144623115 Chr.16: 67436099-67436099 on Build GRCh38
rs951605681 Chr.16: 67436059-67436059 on Build GRCh38
rs1000032379 Chr.16: 67435961-67435961 on Build GRCh38
rs28934592 Chr.16: 67436101-67436101 on Build GRCh38
rs121917780 Chr.16: 67436100-67436100 on Build GRCh38
rs761368643 Chr.16: 67435939-67435939 on Build GRCh38
rs757425144 Chr.16: 67436081-67436081 on Build GRCh38
rs28934591 Chr.16: 67436115-67436115 on Build GRCh38
rs758536896 Chr.16: 67436069-67436069 on Build GRCh38
rs771677502 Chr.16: 67436048-67436048 on Build GRCh38
rs374516226 Chr.16: 67435935-67435935 on Build GRCh38
rs755994842 Chr.16: 67436008-67436008 on Build GRCh38

View Full Product Details