Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145895486-145895223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA10
Entrez Gene ID
8515
Gene Name
integrin subunit alpha 10
Gene Aliases
PRO827
UniGene
Hs.158237
Gene Chromosome Location
Chr.1: 145891208-145910189 on Build GRCh38
Transcripts
NM_001303040.1 NM_001303041.1 NM_003637.4 XM_005277436.3 XM_011510083.2 XM_011510084.1 XM_017002622.1 XM_017002623.1 XM_017002624.1 XM_017002625.1 XM_017002626.1 XM_017002628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4443154 c.3212A>G p.N1071S Chr.1: 145895296-145895296 on Build GRCh38
2120971 c.3138G>T p.Q1046H Chr.1: 145895370-145895370 on Build GRCh38
5377501 c.3222C>T p.F1074F Chr.1: 145895286-145895286 on Build GRCh38
5843463 c.3115-2A>G p.? Chr.1: 145895395-145895395 on Build GRCh38
5353237 c.3196T>C p.L1066L Chr.1: 145895312-145895312 on Build GRCh38
View More

View Full Product Details