Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122729605-122729848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCAR1
Entrez Gene ID
27198
Gene Name
hydroxycarboxylic acid receptor 1
Gene Aliases
FKSG80, GPR104, GPR81, HCA1, LACR1, TA-GPCR, TAGPCR
UniGene
Hs.610873
Gene Chromosome Location
Chr.12: 122727606-122730582 on Build GRCh38
Transcripts
NM_032554.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5042732 c.617G>A p.R206K Chr.12: 122729723-122729723 on Build GRCh38
936310 c.687C>A p.F229L Chr.12: 122729653-122729653 on Build GRCh38
1639063 c.650A>C p.K217T Chr.12: 122729690-122729690 on Build GRCh38
6320217 c.529C>A p.H177N Chr.12: 122729811-122729811 on Build GRCh38
5027710 c.507C>T p.I169I Chr.12: 122729833-122729833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375320732 Chr.12: 122729686-122729686 on Build GRCh38
rs201727220 Chr.12: 122729719-122729719 on Build GRCh38
rs777037364 Chr.12: 122729720-122729720 on Build GRCh38
rs149768068 Chr.12: 122729632-122729632 on Build GRCh38
rs925633077 Chr.12: 122729794-122729794 on Build GRCh38
rs376543632 Chr.12: 122729773-122729773 on Build GRCh38
rs748060410 Chr.12: 122729807-122729807 on Build GRCh38
rs558552970 Chr.12: 122729642-122729642 on Build GRCh38
rs753632848 Chr.12: 122729774-122729774 on Build GRCh38
rs781034960 Chr.12: 122729738-122729741 on Build GRCh38
rs61742326 Chr.12: 122729825-122729825 on Build GRCh38
rs761061194 Chr.12: 122729652-122729652 on Build GRCh38
rs373412840 Chr.12: 122729762-122729762 on Build GRCh38
rs777756646 Chr.12: 122729824-122729824 on Build GRCh38
rs748051752 Chr.12: 122729823-122729828 on Build GRCh38
rs750390654 Chr.12: 122729711-122729711 on Build GRCh38
rs945066304 Chr.12: 122729672-122729672 on Build GRCh38
rs751023135 Chr.12: 122729681-122729681 on Build GRCh38
rs775889828 Chr.12: 122729729-122729729 on Build GRCh38
rs755911344 Chr.12: 122729687-122729687 on Build GRCh38
rs755901720 Chr.12: 122729765-122729765 on Build GRCh38
rs766752596 Chr.12: 122729656-122729656 on Build GRCh38
rs760101875 Chr.12: 122729668-122729668 on Build GRCh38
rs780031221 Chr.12: 122729692-122729692 on Build GRCh38
rs369713712 Chr.12: 122729660-122729660 on Build GRCh38
rs372254985 Chr.12: 122729682-122729682 on Build GRCh38
rs934601804 Chr.12: 122729819-122729819 on Build GRCh38
rs368288293 Chr.12: 122729673-122729673 on Build GRCh38
rs960880928 Chr.12: 122729709-122729709 on Build GRCh38
rs978437931 Chr.12: 122729780-122729780 on Build GRCh38
rs754320989 Chr.12: 122729667-122729667 on Build GRCh38
rs748194436 Chr.12: 122729705-122729705 on Build GRCh38
rs758483547 Chr.12: 122729811-122729811 on Build GRCh38
rs761334856 Chr.12: 122729735-122729735 on Build GRCh38
rs201961889 Chr.12: 122729699-122729699 on Build GRCh38
rs138206647 Chr.12: 122729643-122729643 on Build GRCh38
rs149599946 Chr.12: 122729700-122729700 on Build GRCh38
rs201462968 Chr.12: 122729713-122729713 on Build GRCh38
rs375870381 Chr.12: 122729636-122729636 on Build GRCh38
rs765663615 Chr.12: 122729727-122729727 on Build GRCh38
rs759905684 Chr.12: 122729726-122729726 on Build GRCh38
rs754757000 Chr.12: 122729693-122729694 on Build GRCh38
rs754827402 Chr.12: 122729799-122729799 on Build GRCh38
rs773312705 Chr.12: 122729647-122729647 on Build GRCh38
rs758281047 Chr.12: 122729714-122729714 on Build GRCh38
rs767155988 Chr.12: 122729683-122729683 on Build GRCh38
rs542483050 Chr.12: 122729800-122729800 on Build GRCh38
rs892344961 Chr.12: 122729664-122729664 on Build GRCh38
rs367981732 Chr.12: 122729756-122729756 on Build GRCh38
rs371072988 Chr.12: 122729631-122729631 on Build GRCh38
rs1014802698 Chr.12: 122729767-122729767 on Build GRCh38
rs746721610 Chr.12: 122729685-122729686 on Build GRCh38

View Full Product Details