Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74342997-74343254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF19
Entrez Gene ID
124602
Gene Name
kinesin family member 19
Gene Aliases
KIF19A
UniGene
Hs.372773
Gene Chromosome Location
Chr.17: 74326212-74355820 on Build GRCh38
Transcripts
NM_153209.3 XM_005257029.1 XM_005257030.1 XM_017024150.1 XM_017024151.1 XM_017024152.1 XM_017024153.1 XM_017024154.1 XM_017024155.1 XM_017024156.1 XM_017024157.1 XM_017024158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
984015 c.354C>A p.D118E Chr.17: 74343058-74343058 on Build GRCh38
236145 c.408G>T p.E136D Chr.17: 74343112-74343112 on Build GRCh38
984016 c.438C>A p.V146V Chr.17: 74343142-74343142 on Build GRCh38
6394001 c.359A>T p.E120V Chr.17: 74343063-74343063 on Build GRCh38
2977359 c.435G>T p.E145D Chr.17: 74343139-74343139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750184034 Chr.17: 74343212-74343212 on Build GRCh38
rs564777800 Chr.17: 74343064-74343064 on Build GRCh38
rs752607583 Chr.17: 74343135-74343135 on Build GRCh38
rs201661834 Chr.17: 74343098-74343098 on Build GRCh38
rs758369858 Chr.17: 74343162-74343162 on Build GRCh38
rs575609935 Chr.17: 74343041-74343041 on Build GRCh38
rs748516313 Chr.17: 74343191-74343191 on Build GRCh38
rs777513773 Chr.17: 74343146-74343146 on Build GRCh38
rs747262363 Chr.17: 74343091-74343091 on Build GRCh38
rs376323276 Chr.17: 74343193-74343193 on Build GRCh38
rs376444969 Chr.17: 74343169-74343169 on Build GRCh38
rs751211394 Chr.17: 74343121-74343121 on Build GRCh38
rs762308119 Chr.17: 74343109-74343109 on Build GRCh38
rs148288456 Chr.17: 74343184-74343184 on Build GRCh38
rs779047979 Chr.17: 74343030-74343030 on Build GRCh38
rs756048450 Chr.17: 74343024-74343024 on Build GRCh38
rs772761715 Chr.17: 74343102-74343102 on Build GRCh38
rs767390990 Chr.17: 74343092-74343092 on Build GRCh38
rs141652791 Chr.17: 74343059-74343059 on Build GRCh38
rs373748715 Chr.17: 74343235-74343235 on Build GRCh38
rs781529261 Chr.17: 74343187-74343187 on Build GRCh38
rs746492445 Chr.17: 74343195-74343195 on Build GRCh38
rs1028450575 Chr.17: 74343126-74343126 on Build GRCh38
rs753819350 Chr.17: 74343161-74343161 on Build GRCh38
rs754729242 Chr.17: 74343128-74343128 on Build GRCh38
rs762348071 Chr.17: 74343100-74343100 on Build GRCh38
rs72852205 Chr.17: 74343230-74343230 on Build GRCh38
rs763541480 Chr.17: 74343199-74343199 on Build GRCh38
rs201468425 Chr.17: 74343173-74343173 on Build GRCh38
rs776668350 Chr.17: 74343075-74343089 on Build GRCh38
rs756066443 Chr.17: 74343145-74343145 on Build GRCh38
rs537444829 Chr.17: 74343020-74343020 on Build GRCh38
rs765728283 Chr.17: 74343101-74343101 on Build GRCh38
rs961648695 Chr.17: 74343185-74343185 on Build GRCh38
rs766022152 Chr.17: 74343110-74343110 on Build GRCh38
rs773556087 Chr.17: 74343196-74343196 on Build GRCh38
rs878895606 Chr.17: 74343150-74343150 on Build GRCh38
rs201526020 Chr.17: 74343132-74343132 on Build GRCh38
rs145969437 Chr.17: 74343070-74343070 on Build GRCh38
rs560678311 Chr.17: 74343171-74343171 on Build GRCh38
rs748079343 Chr.17: 74343023-74343023 on Build GRCh38
rs368900515 Chr.17: 74343231-74343231 on Build GRCh38

View Full Product Details