Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 2718056-2718329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNV2
Entrez Gene ID
169522
Gene Name
potassium voltage-gated channel modifier subfamily V member 2
Gene Aliases
KV11.1, Kv8.2, RCD3B
UniGene
Hs.622675
Gene Chromosome Location
Chr.9: 2717526-2730037 on Build GRCh38
Transcripts
NM_133497.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2771492 c.533C>T p.P178L Chr.9: 2718272-2718272 on Build GRCh38
5835725 c.446_447insCTT p.F151_D152insF Chr.9: 2718185-2718186 on Build GRCh38
2771494 c.545T>C p.L182P Chr.9: 2718284-2718284 on Build GRCh38
4499487 c.543C>T p.F181F Chr.9: 2718282-2718282 on Build GRCh38
5876089 c.354_355delCC p.P119fs*252 Chr.9: 2718093-2718094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755292064 Chr.9: 2718146-2718146 on Build GRCh38
rs746165503 Chr.9: 2718239-2718239 on Build GRCh38
rs530006522 Chr.9: 2718171-2718171 on Build GRCh38
rs759241107 Chr.9: 2718118-2718118 on Build GRCh38
rs773293657 Chr.9: 2718253-2718253 on Build GRCh38
rs1019407895 Chr.9: 2718229-2718229 on Build GRCh38
rs563575183 Chr.9: 2718209-2718209 on Build GRCh38
rs752649092 Chr.9: 2718190-2718190 on Build GRCh38
rs754619195 Chr.9: 2718294-2718294 on Build GRCh38
rs886131960 Chr.9: 2718159-2718159 on Build GRCh38
rs767584771 Chr.9: 2718217-2718217 on Build GRCh38
rs762556896 Chr.9: 2718174-2718174 on Build GRCh38
rs777051584 Chr.9: 2718169-2718169 on Build GRCh38
rs370938953 Chr.9: 2718204-2718204 on Build GRCh38
rs932944724 Chr.9: 2718150-2718150 on Build GRCh38
rs768859361 Chr.9: 2718312-2718312 on Build GRCh38
rs773801117 Chr.9: 2718208-2718208 on Build GRCh38
rs531689254 Chr.9: 2718252-2718252 on Build GRCh38
rs140256288 Chr.9: 2718181-2718181 on Build GRCh38
rs397514604 Chr.9: 2718230-2718230 on Build GRCh38
rs765877654 Chr.9: 2718106-2718106 on Build GRCh38
rs746257061 Chr.9: 2718197-2718197 on Build GRCh38
rs764199468 Chr.9: 2718231-2718231 on Build GRCh38
rs781689544 Chr.9: 2718237-2718237 on Build GRCh38
rs772921412 Chr.9: 2718303-2718303 on Build GRCh38
rs772682207 Chr.9: 2718101-2718101 on Build GRCh38
rs143936775 Chr.9: 2718130-2718130 on Build GRCh38
rs767955052 Chr.9: 2718275-2718275 on Build GRCh38
rs778432513 Chr.9: 2718295-2718295 on Build GRCh38
rs751274302 Chr.9: 2718135-2718135 on Build GRCh38
rs758516182 Chr.9: 2718192-2718192 on Build GRCh38
rs766187569 Chr.9: 2718262-2718262 on Build GRCh38
rs915410588 Chr.9: 2718093-2718093 on Build GRCh38
rs104894113 Chr.9: 2718166-2718166 on Build GRCh38
rs748571527 Chr.9: 2718094-2718094 on Build GRCh38
rs202166642 Chr.9: 2718084-2718084 on Build GRCh38
rs747275875 Chr.9: 2718164-2718164 on Build GRCh38
rs201520619 Chr.9: 2718259-2718259 on Build GRCh38
rs759283660 Chr.9: 2718308-2718308 on Build GRCh38
rs779113871 Chr.9: 2718151-2718151 on Build GRCh38
rs202036979 Chr.9: 2718078-2718078 on Build GRCh38
rs778345203 Chr.9: 2718157-2718157 on Build GRCh38
rs766337592 Chr.9: 2718206-2718206 on Build GRCh38
rs760548669 Chr.9: 2718220-2718220 on Build GRCh38
rs1004802068 Chr.9: 2718161-2718161 on Build GRCh38
rs977790637 Chr.9: 2718301-2718301 on Build GRCh38
rs761130319 Chr.9: 2718103-2718103 on Build GRCh38
rs886408864 Chr.9: 2718198-2718198 on Build GRCh38
rs149893780 Chr.9: 2718224-2718224 on Build GRCh38
rs758432224 Chr.9: 2718077-2718077 on Build GRCh38
rs200711754 Chr.9: 2718203-2718203 on Build GRCh38
rs773398432 Chr.9: 2718102-2718102 on Build GRCh38
rs751529297 Chr.9: 2718178-2718178 on Build GRCh38
rs200662596 Chr.9: 2718274-2718274 on Build GRCh38
rs751644579 Chr.9: 2718233-2718233 on Build GRCh38
rs756486876 Chr.9: 2718240-2718240 on Build GRCh38
rs144949136 Chr.9: 2718272-2718272 on Build GRCh38
rs953979254 Chr.9: 2718149-2718149 on Build GRCh38
rs770819821 Chr.9: 2718304-2718304 on Build GRCh38
rs780435673 Chr.9: 2718292-2718292 on Build GRCh38
rs751709716 Chr.9: 2718277-2718277 on Build GRCh38
rs200353727 Chr.9: 2718088-2718088 on Build GRCh38
rs768870558 Chr.9: 2718085-2718085 on Build GRCh38
rs764727253 Chr.9: 2718127-2718127 on Build GRCh38
rs776442018 Chr.9: 2718306-2718306 on Build GRCh38
rs771471592 Chr.9: 2718196-2718196 on Build GRCh38
rs761713749 Chr.9: 2718179-2718179 on Build GRCh38
rs933040040 Chr.9: 2718245-2718245 on Build GRCh38
rs141099767 Chr.9: 2718118-2718119 on Build GRCh38
rs373196631 Chr.9: 2718138-2718138 on Build GRCh38
rs757715024 Chr.9: 2718234-2718234 on Build GRCh38
rs751600925 Chr.9: 2718268-2718268 on Build GRCh38
rs377667539 Chr.9: 2718193-2718193 on Build GRCh38
rs771269109 Chr.9: 2718167-2718167 on Build GRCh38
rs547964930 Chr.9: 2718297-2718297 on Build GRCh38
rs199735148 Chr.9: 2718137-2718137 on Build GRCh38
rs753795579 Chr.9: 2718267-2718273 on Build GRCh38
rs769400306 Chr.9: 2718199-2718199 on Build GRCh38
rs760750739 Chr.9: 2718256-2718256 on Build GRCh38
rs541196988 Chr.9: 2718128-2718128 on Build GRCh38
rs754099727 Chr.9: 2718144-2718144 on Build GRCh38
rs765596301 Chr.9: 2718165-2718165 on Build GRCh38
rs746873819 Chr.9: 2718079-2718079 on Build GRCh38
rs776275880 Chr.9: 2718116-2718116 on Build GRCh38
rs747690555 Chr.9: 2718249-2718249 on Build GRCh38
rs774816084 Chr.9: 2718172-2718172 on Build GRCh38
rs780340984 Chr.9: 2718241-2718241 on Build GRCh38
rs763872477 Chr.9: 2718176-2718176 on Build GRCh38
rs762314332 Chr.9: 2718205-2718205 on Build GRCh38
rs748966973 Chr.9: 2718242-2718242 on Build GRCh38
rs1023410136 Chr.9: 2718290-2718290 on Build GRCh38
rs748280472 Chr.9: 2718156-2718156 on Build GRCh38
rs764249096 Chr.9: 2718265-2718265 on Build GRCh38
rs752621455 Chr.9: 2718076-2718076 on Build GRCh38
rs146659188 Chr.9: 2718180-2718180 on Build GRCh38
rs201674494 Chr.9: 2718083-2718083 on Build GRCh38
rs1012003891 Chr.9: 2718289-2718289 on Build GRCh38
rs773099220 Chr.9: 2718219-2718219 on Build GRCh38
rs766549271 Chr.9: 2718186-2718188 on Build GRCh38
rs370129493 Chr.9: 2718139-2718139 on Build GRCh38
rs750918937 Chr.9: 2718278-2718278 on Build GRCh38
rs771691876 Chr.9: 2718298-2718298 on Build GRCh38
rs534069865 Chr.9: 2718280-2718280 on Build GRCh38
rs945133207 Chr.9: 2718170-2718170 on Build GRCh38
rs915176607 Chr.9: 2718281-2718281 on Build GRCh38
rs376320246 Chr.9: 2718264-2718264 on Build GRCh38
rs768242305 Chr.9: 2718244-2718244 on Build GRCh38
rs747542620 Chr.9: 2718195-2718195 on Build GRCh38

View Full Product Details