Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113655400-113655657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USF3
Entrez Gene ID
205717
Gene Name
upstream transcription factor family member 3
Gene Aliases
KIAA2018
UniGene
Hs.632570
Gene Chromosome Location
Chr.3: 113648385-113696657 on Build GRCh38
Transcripts
NM_001009899.3 NR_111981.1 XM_005247208.4 XM_017005871.1 XM_017005872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4112334 c.6277A>G p.T2093A Chr.3: 113655405-113655405 on Build GRCh38
6502461 c.6155C>T p.S2052L Chr.3: 113655527-113655527 on Build GRCh38
5974487 c.6205C>T p.P2069S Chr.3: 113655477-113655477 on Build GRCh38
4386815 c.6199T>C p.F2067L Chr.3: 113655483-113655483 on Build GRCh38
5716028 c.6120C>T p.F2040F Chr.3: 113655562-113655562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868750493 Chr.3: 113655608-113655608 on Build GRCh38
rs746350716 Chr.3: 113655479-113655479 on Build GRCh38
rs149001610 Chr.3: 113655481-113655481 on Build GRCh38
rs746737778 Chr.3: 113655634-113655634 on Build GRCh38
rs774007402 Chr.3: 113655519-113655519 on Build GRCh38
rs769879030 Chr.3: 113655596-113655597 on Build GRCh38
rs778111846 Chr.3: 113655597-113655598 on Build GRCh38
rs771869355 Chr.3: 113655535-113655535 on Build GRCh38
rs555995730 Chr.3: 113655550-113655550 on Build GRCh38
rs748831747 Chr.3: 113655603-113655603 on Build GRCh38
rs372772175 Chr.3: 113655467-113655467 on Build GRCh38
rs765685583 Chr.3: 113655561-113655561 on Build GRCh38
rs750963413 Chr.3: 113655590-113655590 on Build GRCh38
rs781576801 Chr.3: 113655470-113655470 on Build GRCh38
rs373111363 Chr.3: 113655630-113655630 on Build GRCh38
rs778471296 Chr.3: 113655606-113655606 on Build GRCh38
rs376015347 Chr.3: 113655507-113655507 on Build GRCh38
rs74980526 Chr.3: 113655562-113655562 on Build GRCh38
rs755561773 Chr.3: 113655509-113655509 on Build GRCh38
rs765366670 Chr.3: 113655551-113655551 on Build GRCh38
rs369738835 Chr.3: 113655567-113655567 on Build GRCh38
rs368472959 Chr.3: 113655460-113655460 on Build GRCh38
rs377319556 Chr.3: 113655566-113655566 on Build GRCh38
rs374781281 Chr.3: 113655539-113655539 on Build GRCh38
rs201490294 Chr.3: 113655598-113655598 on Build GRCh38
rs373643148 Chr.3: 113655584-113655584 on Build GRCh38
rs377443949 Chr.3: 113655636-113655636 on Build GRCh38
rs1043669360 Chr.3: 113655476-113655476 on Build GRCh38
rs752973097 Chr.3: 113655429-113655429 on Build GRCh38
rs746855846 Chr.3: 113655539-113655547 on Build GRCh38
rs1007316850 Chr.3: 113655597-113655597 on Build GRCh38
rs777902214 Chr.3: 113655456-113655456 on Build GRCh38
rs779710277 Chr.3: 113655505-113655505 on Build GRCh38
rs767417630 Chr.3: 113655605-113655605 on Build GRCh38
rs369864905 Chr.3: 113655508-113655508 on Build GRCh38
rs867970188 Chr.3: 113655472-113655472 on Build GRCh38
rs371204248 Chr.3: 113655527-113655527 on Build GRCh38
rs765278691 Chr.3: 113655425-113655425 on Build GRCh38
rs1052381254 Chr.3: 113655439-113655439 on Build GRCh38
rs369155033 Chr.3: 113655547-113655547 on Build GRCh38
rs11540067 Chr.3: 113655513-113655513 on Build GRCh38
rs375100132 Chr.3: 113655437-113655437 on Build GRCh38
rs866066509 Chr.3: 113655491-113655491 on Build GRCh38
rs542739936 Chr.3: 113655581-113655581 on Build GRCh38
rs759575786 Chr.3: 113655424-113655424 on Build GRCh38
rs747815590 Chr.3: 113655626-113655626 on Build GRCh38
rs371309418 Chr.3: 113655627-113655627 on Build GRCh38

View Full Product Details