Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83309267-83309506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPSE
Entrez Gene ID
10855
Gene Name
heparanase
Gene Aliases
HPA, HPA1, HPR1, HPSE1, HSE1
UniGene
Hs.44227
Gene Chromosome Location
Chr.4: 83292461-83335153 on Build GRCh38
Transcripts
NM_001098540.2 NM_001166498.2 NM_001199830.1 NM_006665.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418572 c.975A>T p.K325N Chr.4: 83309411-83309411 on Build GRCh38
1131015 c.920A>G p.K307R Chr.4: 83309466-83309466 on Build GRCh38
4126385 c.965C>T p.S322F Chr.4: 83309421-83309421 on Build GRCh38
3606594 c.921G>A p.K307K Chr.4: 83309465-83309465 on Build GRCh38
3606593 c.982C>T p.Q328* Chr.4: 83309404-83309404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138290665 Chr.4: 83309459-83309459 on Build GRCh38
rs779763952 Chr.4: 83309449-83309449 on Build GRCh38
rs755661805 Chr.4: 83309440-83309440 on Build GRCh38
rs771052437 Chr.4: 83309383-83309383 on Build GRCh38
rs750073134 Chr.4: 83309438-83309438 on Build GRCh38
rs769619708 Chr.4: 83309395-83309395 on Build GRCh38
rs924964996 Chr.4: 83309296-83309296 on Build GRCh38
rs747045851 Chr.4: 83309377-83309377 on Build GRCh38
rs11099592 Chr.4: 83309466-83309466 on Build GRCh38
rs552720199 Chr.4: 83309394-83309394 on Build GRCh38
rs776881606 Chr.4: 83309385-83309385 on Build GRCh38
rs775574185 Chr.4: 83309301-83309301 on Build GRCh38
rs764072489 Chr.4: 83309402-83309402 on Build GRCh38
rs751596196 Chr.4: 83309412-83309412 on Build GRCh38
rs760424522 Chr.4: 83309411-83309411 on Build GRCh38
rs761372448 Chr.4: 83309416-83309416 on Build GRCh38
rs764024455 Chr.4: 83309451-83309451 on Build GRCh38
rs766999252 Chr.4: 83309417-83309417 on Build GRCh38
rs200961522 Chr.4: 83309401-83309401 on Build GRCh38
rs867060728 Chr.4: 83309365-83309365 on Build GRCh38
rs186085700 Chr.4: 83309451-83309451 on Build GRCh38
rs758787707 Chr.4: 83309455-83309455 on Build GRCh38
rs1054785512 Chr.4: 83309308-83309308 on Build GRCh38
rs1035952283 Chr.4: 83309398-83309398 on Build GRCh38
rs530939016 Chr.4: 83309327-83309327 on Build GRCh38

View Full Product Details