Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82055059-82055326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPS1
Entrez Gene ID
2873
Gene Name
G protein pathway suppressor 1
Gene Aliases
COPS1, CSN1, SGN1
UniGene
Hs.268530
Gene Chromosome Location
Chr.17: 82051477-82057470 on Build GRCh38
Transcripts
NM_001321089.1 NM_001321090.1 NM_001321091.1 NM_001321092.1 NM_001321093.1 NM_004127.5 NM_212492.2 XM_005256350.1 XM_005256353.1 XM_005256359.1 XM_005256360.3 XM_011523569.1 XM_011523570.1 XM_017024524.1 XM_017024525.1 XM_017024526.1 XM_017024527.1 XM_017024528.1 XM_017024529.1 XM_017024530.1 XM_017024531.1 XM_017024532.1 XM_017024533.1 XM_017024534.1 XM_017024535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3691950 c.866C>T p.A289V Chr.17: 82055220-82055220 on Build GRCh38
5446505 c.852_853insAC p.L285fs*101 Chr.17: 82055206-82055207 on Build GRCh38
234762 c.819G>A p.E273E Chr.17: 82055173-82055173 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764030794 Chr.17: 82055182-82055182 on Build GRCh38
rs765958479 Chr.17: 82055102-82055102 on Build GRCh38
rs1045192727 Chr.17: 82055240-82055240 on Build GRCh38
rs771228447 Chr.17: 82055085-82055086 on Build GRCh38
rs553675736 Chr.17: 82055128-82055128 on Build GRCh38
rs544740357 Chr.17: 82055173-82055173 on Build GRCh38
rs759396683 Chr.17: 82055108-82055108 on Build GRCh38
rs1028835373 Chr.17: 82055304-82055304 on Build GRCh38
rs770112649 Chr.17: 82055131-82055131 on Build GRCh38
rs1004828423 Chr.17: 82055281-82055281 on Build GRCh38
rs574840143 Chr.17: 82055191-82055191 on Build GRCh38
rs1056667649 Chr.17: 82055232-82055232 on Build GRCh38
rs776091017 Chr.17: 82055295-82055295 on Build GRCh38
rs1012367353 Chr.17: 82055090-82055090 on Build GRCh38
rs950631699 Chr.17: 82055112-82055112 on Build GRCh38
rs368926784 Chr.17: 82055124-82055124 on Build GRCh38
rs746081727 Chr.17: 82055130-82055130 on Build GRCh38
rs896476042 Chr.17: 82055084-82055084 on Build GRCh38
rs983526986 Chr.17: 82055116-82055116 on Build GRCh38
rs780437451 Chr.17: 82055142-82055142 on Build GRCh38
rs11539402 Chr.17: 82055188-82055188 on Build GRCh38
rs762817268 Chr.17: 82055179-82055179 on Build GRCh38
rs776326946 Chr.17: 82055158-82055158 on Build GRCh38
rs554571345 Chr.17: 82055093-82055093 on Build GRCh38
rs772723271 Chr.17: 82055148-82055148 on Build GRCh38
rs948032450 Chr.17: 82055228-82055228 on Build GRCh38
rs946713639 Chr.17: 82055126-82055126 on Build GRCh38
rs755445171 Chr.17: 82055261-82055261 on Build GRCh38
rs749792698 Chr.17: 82055144-82055144 on Build GRCh38
rs758323318 Chr.17: 82055114-82055114 on Build GRCh38
rs760115971 Chr.17: 82055149-82055149 on Build GRCh38
rs772739516 Chr.17: 82055094-82055094 on Build GRCh38
rs567194030 Chr.17: 82055099-82055099 on Build GRCh38
rs965757639 Chr.17: 82055302-82055302 on Build GRCh38
rs780306335 Chr.17: 82055250-82055250 on Build GRCh38
rs770427061 Chr.17: 82055155-82055155 on Build GRCh38
rs750722265 Chr.17: 82055220-82055220 on Build GRCh38
rs746100987 Chr.17: 82055281-82055282 on Build GRCh38
rs371801171 Chr.17: 82055256-82055256 on Build GRCh38
rs35085155 Chr.17: 82055121-82055121 on Build GRCh38
rs368410006 Chr.17: 82055105-82055105 on Build GRCh38
rs765071154 Chr.17: 82055109-82055109 on Build GRCh38
rs767691039 Chr.17: 82055211-82055211 on Build GRCh38
rs898555273 Chr.17: 82055255-82055255 on Build GRCh38
rs369253146 Chr.17: 82055218-82055218 on Build GRCh38
rs777742756 Chr.17: 82055120-82055120 on Build GRCh38
rs372173472 Chr.17: 82055111-82055111 on Build GRCh38
rs756320575 Chr.17: 82055239-82055239 on Build GRCh38
rs759169597 Chr.17: 82055166-82055166 on Build GRCh38
rs566855360 Chr.17: 82055089-82055090 on Build GRCh38
rs764978700 Chr.17: 82055171-82055171 on Build GRCh38
rs543932597 Chr.17: 82055275-82055275 on Build GRCh38
rs749981722 Chr.17: 82055141-82055141 on Build GRCh38
rs1027452291 Chr.17: 82055266-82055266 on Build GRCh38
rs757566285 Chr.17: 82055122-82055122 on Build GRCh38
rs769157899 Chr.17: 82055145-82055145 on Build GRCh38
rs779466369 Chr.17: 82055270-82055270 on Build GRCh38
rs966084458 Chr.17: 82055140-82055140 on Build GRCh38
rs541840696 Chr.17: 82055283-82055283 on Build GRCh38
rs775312656 Chr.17: 82055175-82055175 on Build GRCh38

View Full Product Details