Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162371612-162371881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH7
Entrez Gene ID
90134
Gene Name
potassium voltage-gated channel subfamily H member 7
Gene Aliases
ERG3, HERG3, Kv11.3
UniGene
Hs.657413
Gene Chromosome Location
Chr.2: 162371407-162838747 on Build GRCh38
Transcripts
NM_033272.3 XM_011512109.2 XM_017005218.1 XM_017005219.1 XM_017005220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3568538 c.3539G>A p.G1180E Chr.2: 162371881-162371881 on Build GRCh38
5922167 c.3584G>A p.G1195E Chr.2: 162371836-162371836 on Build GRCh38
4086026 c.3551T>G p.L1184R Chr.2: 162371869-162371869 on Build GRCh38
4582750 c.3567T>C p.S1189S Chr.2: 162371853-162371853 on Build GRCh38
3938577 c.3554A>G p.H1185R Chr.2: 162371866-162371866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534785053 Chr.2: 162371659-162371659 on Build GRCh38
rs770713731 Chr.2: 162371729-162371729 on Build GRCh38
rs1057508923 Chr.2: 162371734-162371734 on Build GRCh38
rs1005044259 Chr.2: 162371817-162371817 on Build GRCh38
rs779617348 Chr.2: 162371791-162371791 on Build GRCh38
rs969305335 Chr.2: 162371765-162371765 on Build GRCh38
rs764940868 Chr.2: 162371803-162371803 on Build GRCh38
rs1010515194 Chr.2: 162371722-162371722 on Build GRCh38
rs956010553 Chr.2: 162371720-162371720 on Build GRCh38
rs771839703 Chr.2: 162371642-162371642 on Build GRCh38
rs187984820 Chr.2: 162371839-162371839 on Build GRCh38
rs557313177 Chr.2: 162371641-162371641 on Build GRCh38
rs1030372118 Chr.2: 162371694-162371694 on Build GRCh38
rs750125664 Chr.2: 162371804-162371804 on Build GRCh38
rs753432481 Chr.2: 162371783-162371783 on Build GRCh38
rs373993413 Chr.2: 162371814-162371814 on Build GRCh38
rs755000626 Chr.2: 162371838-162371838 on Build GRCh38
rs183509596 Chr.2: 162371728-162371728 on Build GRCh38
rs774496305 Chr.2: 162371852-162371852 on Build GRCh38
rs771019746 Chr.2: 162371850-162371850 on Build GRCh38
rs777608961 Chr.2: 162371670-162371670 on Build GRCh38
rs947449815 Chr.2: 162371716-162371716 on Build GRCh38
rs201273498 Chr.2: 162371767-162371767 on Build GRCh38
rs756827076 Chr.2: 162371793-162371793 on Build GRCh38
rs980257719 Chr.2: 162371781-162371781 on Build GRCh38
rs913353313 Chr.2: 162371683-162371683 on Build GRCh38
rs749426625 Chr.2: 162371847-162371847 on Build GRCh38
rs776649582 Chr.2: 162371810-162371810 on Build GRCh38
rs150975356 Chr.2: 162371778-162371779 on Build GRCh38
rs918278123 Chr.2: 162371853-162371853 on Build GRCh38
rs746917781 Chr.2: 162371834-162371834 on Build GRCh38

View Full Product Details