Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16154306-16154571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSH2D
Entrez Gene ID
84941
Gene Name
hematopoietic SH2 domain containing
Gene Aliases
ALX, HSH2
UniGene
Hs.631617
Gene Chromosome Location
Chr.19: 16134028-16158575 on Build GRCh38
Transcripts
NM_001291274.1 NM_032855.3 NR_111903.1 NR_111904.1 XM_011528386.2 XM_011528387.2 XM_011528388.2 XM_011528389.2 XM_011528390.1 XM_017027402.1 XM_017027403.1 XM_017027404.1

SNP Detail

SNP ID Chromosome Location
rs1014934871 Chr.19: 16154446-16154446 on Build GRCh38
rs367693992 Chr.19: 16154394-16154394 on Build GRCh38
rs879576560 Chr.19: 16154365-16154365 on Build GRCh38
rs755161694 Chr.19: 16154390-16154390 on Build GRCh38
rs773692980 Chr.19: 16154491-16154491 on Build GRCh38
rs903174575 Chr.19: 16154472-16154472 on Build GRCh38
rs771962827 Chr.19: 16154522-16154522 on Build GRCh38
rs1021616691 Chr.19: 16154328-16154328 on Build GRCh38
rs35467188 Chr.19: 16154370-16154370 on Build GRCh38
rs367861665 Chr.19: 16154455-16154455 on Build GRCh38
rs754083096 Chr.19: 16154356-16154356 on Build GRCh38
rs892920227 Chr.19: 16154481-16154481 on Build GRCh38
rs758768377 Chr.19: 16154415-16154415 on Build GRCh38
rs756754513 Chr.19: 16154393-16154393 on Build GRCh38
rs761813062 Chr.19: 16154510-16154510 on Build GRCh38
rs371673671 Chr.19: 16154473-16154473 on Build GRCh38
rs752087568 Chr.19: 16154386-16154386 on Build GRCh38
rs376039892 Chr.19: 16154422-16154422 on Build GRCh38
rs768234036 Chr.19: 16154480-16154480 on Build GRCh38
rs972768962 Chr.19: 16154398-16154398 on Build GRCh38
rs1003644184 Chr.19: 16154341-16154341 on Build GRCh38
rs746360628 Chr.19: 16154550-16154550 on Build GRCh38
rs990847918 Chr.19: 16154385-16154385 on Build GRCh38
rs1056371873 Chr.19: 16154429-16154429 on Build GRCh38
rs36088948 Chr.19: 16154431-16154431 on Build GRCh38
rs918938694 Chr.19: 16154389-16154389 on Build GRCh38
rs942533396 Chr.19: 16154445-16154445 on Build GRCh38
rs957199697 Chr.19: 16154326-16154326 on Build GRCh38
rs537189088 Chr.19: 16154449-16154449 on Build GRCh38
rs1046012106 Chr.19: 16154464-16154464 on Build GRCh38
rs555553292 Chr.19: 16154414-16154414 on Build GRCh38
rs377096035 Chr.19: 16154434-16154434 on Build GRCh38
rs368588071 Chr.19: 16154423-16154423 on Build GRCh38
rs376371671 Chr.19: 16154407-16154407 on Build GRCh38
rs190808280 Chr.19: 16154401-16154401 on Build GRCh38
rs117103057 Chr.19: 16154352-16154352 on Build GRCh38
rs376245483 Chr.19: 16154531-16154531 on Build GRCh38
rs78876877 Chr.19: 16154334-16154334 on Build GRCh38

View Full Product Details