Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 51384652-51384913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISL1
Entrez Gene ID
3670
Gene Name
ISL LIM homeobox 1
Gene Aliases
ISLET1, Isl-1
UniGene
Hs.505
Gene Chromosome Location
Chr.5: 51383124-51394730 on Build GRCh38
Transcripts
NM_002202.2 XM_011543380.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3343239 c.162G>A p.E54E Chr.5: 51384674-51384674 on Build GRCh38
379978 c.170C>T p.T57I Chr.5: 51384682-51384682 on Build GRCh38
5436716 c.160G>A p.E54K Chr.5: 51384672-51384672 on Build GRCh38
738616 c.189G>A p.G63G Chr.5: 51384701-51384701 on Build GRCh38
3429495 c.206G>T p.R69I Chr.5: 51384718-51384718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769805341 Chr.5: 51384780-51384780 on Build GRCh38
rs535686797 Chr.5: 51384827-51384827 on Build GRCh38
rs151276558 Chr.5: 51384874-51384875 on Build GRCh38
rs938032865 Chr.5: 51384813-51384813 on Build GRCh38
rs926648468 Chr.5: 51384737-51384737 on Build GRCh38
rs753242388 Chr.5: 51384709-51384709 on Build GRCh38
rs747176332 Chr.5: 51384707-51384707 on Build GRCh38
rs1056910531 Chr.5: 51384868-51384868 on Build GRCh38
rs777485181 Chr.5: 51384701-51384701 on Build GRCh38
rs758205204 Chr.5: 51384692-51384692 on Build GRCh38
rs746803234 Chr.5: 51384746-51384749 on Build GRCh38
rs566999697 Chr.5: 51384795-51384795 on Build GRCh38
rs374264309 Chr.5: 51384750-51384750 on Build GRCh38

View Full Product Details