Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133043689-133043960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C5
Entrez Gene ID
9328
Gene Name
general transcription factor IIIC subunit 5
Gene Aliases
TFIIIC63, TFIIICepsilon, TFiiiC2-63
UniGene
Hs.495417
Gene Chromosome Location
Chr.9: 133030675-133058503 on Build GRCh38
Transcripts
NM_001122823.1 NM_001286709.1 NM_012087.3 XM_005272234.3 XM_011519201.1 XM_017015315.1 XM_017015316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4689206 c.463G>A p.E155K Chr.9: 133043817-133043817 on Build GRCh38
4614080 c.129delC p.P45fs*35 Chr.9: 133043858-133043858 on Build GRCh38
1643767 c.407C>T p.T136M Chr.9: 133043761-133043761 on Build GRCh38
6676850 c.303G>T p.K101N Chr.9: 133043798-133043798 on Build GRCh38
4649291 c.406A>G p.T136A Chr.9: 133043760-133043760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527961085 Chr.9: 133043938-133043938 on Build GRCh38
rs768655384 Chr.9: 133043795-133043795 on Build GRCh38
rs774324292 Chr.9: 133043856-133043856 on Build GRCh38
rs139422900 Chr.9: 133043812-133043812 on Build GRCh38
rs760341869 Chr.9: 133043857-133043858 on Build GRCh38
rs976486579 Chr.9: 133043879-133043879 on Build GRCh38
rs749793500 Chr.9: 133043787-133043787 on Build GRCh38
rs369628277 Chr.9: 133043816-133043816 on Build GRCh38
rs769157658 Chr.9: 133043855-133043855 on Build GRCh38
rs762062722 Chr.9: 133043719-133043719 on Build GRCh38
rs777306828 Chr.9: 133043768-133043768 on Build GRCh38
rs140794229 Chr.9: 133043873-133043873 on Build GRCh38
rs781716105 Chr.9: 133043775-133043775 on Build GRCh38
rs147270006 Chr.9: 133043730-133043730 on Build GRCh38
rs752569050 Chr.9: 133043937-133043938 on Build GRCh38
rs745499137 Chr.9: 133043842-133043842 on Build GRCh38
rs775157297 Chr.9: 133043937-133043937 on Build GRCh38
rs774466659 Chr.9: 133043802-133043802 on Build GRCh38
rs746171889 Chr.9: 133043782-133043782 on Build GRCh38
rs151291681 Chr.9: 133043810-133043810 on Build GRCh38
rs200990519 Chr.9: 133043805-133043805 on Build GRCh38
rs766210383 Chr.9: 133043725-133043725 on Build GRCh38
rs36017796 Chr.9: 133043891-133043891 on Build GRCh38
rs763539610 Chr.9: 133043817-133043817 on Build GRCh38
rs138342612 Chr.9: 133043849-133043849 on Build GRCh38
rs202102841 Chr.9: 133043886-133043886 on Build GRCh38
rs777809658 Chr.9: 133043926-133043926 on Build GRCh38
rs200449975 Chr.9: 133043936-133043936 on Build GRCh38
rs150454891 Chr.9: 133043783-133043783 on Build GRCh38
rs756609941 Chr.9: 133043843-133043843 on Build GRCh38
rs897524693 Chr.9: 133043742-133043742 on Build GRCh38
rs765102902 Chr.9: 133043753-133043753 on Build GRCh38
rs750117418 Chr.9: 133043716-133043717 on Build GRCh38
rs755803250 Chr.9: 133043905-133043905 on Build GRCh38
rs760162906 Chr.9: 133043732-133043732 on Build GRCh38
rs932025853 Chr.9: 133043887-133043887 on Build GRCh38
rs573014629 Chr.9: 133043804-133043804 on Build GRCh38
rs189383196 Chr.9: 133043731-133043731 on Build GRCh38
rs144724577 Chr.9: 133043877-133043877 on Build GRCh38
rs764216043 Chr.9: 133043714-133043714 on Build GRCh38
rs897130126 Chr.9: 133043741-133043741 on Build GRCh38
rs751042827 Chr.9: 133043824-133043824 on Build GRCh38
rs764314909 Chr.9: 133043761-133043761 on Build GRCh38
rs575422131 Chr.9: 133043762-133043762 on Build GRCh38
rs554887635 Chr.9: 133043786-133043786 on Build GRCh38
rs775308894 Chr.9: 133043878-133043878 on Build GRCh38
rs772179043 Chr.9: 133043924-133043924 on Build GRCh38
rs1051109097 Chr.9: 133043779-133043779 on Build GRCh38
rs754421900 Chr.9: 133043905-133043906 on Build GRCh38
rs34305524 Chr.9: 133043902-133043902 on Build GRCh38
rs778947914 Chr.9: 133043933-133043935 on Build GRCh38
rs753342533 Chr.9: 133043886-133043887 on Build GRCh38
rs754188625 Chr.9: 133043908-133043908 on Build GRCh38
rs766020367 Chr.9: 133043869-133043871 on Build GRCh38
rs376483496 Chr.9: 133043863-133043863 on Build GRCh38
rs898070022 Chr.9: 133043933-133043933 on Build GRCh38
rs750497049 Chr.9: 133043867-133043867 on Build GRCh38
rs780302406 Chr.9: 133043907-133043907 on Build GRCh38
rs747256128 Chr.9: 133043933-133043933 on Build GRCh38
rs140767076 Chr.9: 133043771-133043771 on Build GRCh38
rs376409247 Chr.9: 133043898-133043898 on Build GRCh38
rs751047660 Chr.9: 133043890-133043890 on Build GRCh38
rs150056568 Chr.9: 133043838-133043838 on Build GRCh38
rs772127575 Chr.9: 133043865-133043865 on Build GRCh38
rs774499435 Chr.9: 133043716-133043716 on Build GRCh38
rs370278139 Chr.9: 133043918-133043918 on Build GRCh38
rs1033571304 Chr.9: 133043792-133043792 on Build GRCh38
rs754914868 Chr.9: 133043927-133043927 on Build GRCh38
rs756977966 Chr.9: 133043837-133043837 on Build GRCh38
rs1047844 Chr.9: 133043785-133043785 on Build GRCh38
rs748650772 Chr.9: 133043922-133043922 on Build GRCh38
rs756925114 Chr.9: 133043772-133043772 on Build GRCh38
rs368008906 Chr.9: 133043848-133043848 on Build GRCh38
rs370716957 Chr.9: 133043885-133043885 on Build GRCh38
rs181363313 Chr.9: 133043811-133043811 on Build GRCh38

View Full Product Details