Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46689494-46689749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB14
Entrez Gene ID
9813
Gene Name
EF-hand calcium binding domain 14
Gene Aliases
KIAA0494
UniGene
Hs.707688
Gene Chromosome Location
Chr.1: 46675159-46719064 on Build GRCh38
Transcripts
NM_014774.2 NR_038827.1 NR_038828.1

Gene Symbol
EFCAB14-AS1
Entrez Gene ID
100130197
Gene Name
EFCAB14 antisense RNA 1
Gene Aliases
KIAA0494-AS1
UniGene
Hs.568643
Gene Chromosome Location
Chr.1: 46674036-46692098 on Build GRCh38
Transcripts
NM_014774.2 NR_038827.1 NR_038828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3490344 c.789G>A p.L263L Chr.1: 46689593-46689593 on Build GRCh38
3490345 c.732C>T p.I244I Chr.1: 46689650-46689650 on Build GRCh38
6651857 c.737C>T p.P246L Chr.1: 46689645-46689645 on Build GRCh38
6651859 c.795+9_795+10insT p.? Chr.1: 46689577-46689578 on Build GRCh38
4008609 c.772A>G p.T258A Chr.1: 46689610-46689610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778374712 Chr.1: 46689620-46689620 on Build GRCh38
rs993941497 Chr.1: 46689574-46689574 on Build GRCh38
rs775704231 Chr.1: 46689712-46689712 on Build GRCh38
rs200564580 Chr.1: 46689619-46689619 on Build GRCh38
rs770789971 Chr.1: 46689650-46689650 on Build GRCh38
rs979527765 Chr.1: 46689520-46689520 on Build GRCh38
rs1029479107 Chr.1: 46689615-46689615 on Build GRCh38
rs897115452 Chr.1: 46689529-46689529 on Build GRCh38
rs750211749 Chr.1: 46689700-46689701 on Build GRCh38
rs371888407 Chr.1: 46689582-46689582 on Build GRCh38
rs919659505 Chr.1: 46689629-46689629 on Build GRCh38
rs781506952 Chr.1: 46689606-46689606 on Build GRCh38
rs753477515 Chr.1: 46689604-46689605 on Build GRCh38
rs956737940 Chr.1: 46689533-46689533 on Build GRCh38
rs763125940 Chr.1: 46689715-46689715 on Build GRCh38
rs1018361695 Chr.1: 46689687-46689687 on Build GRCh38
rs751902287 Chr.1: 46689590-46689590 on Build GRCh38
rs747716183 Chr.1: 46689638-46689638 on Build GRCh38
rs778913667 Chr.1: 46689695-46689695 on Build GRCh38
rs947872045 Chr.1: 46689618-46689618 on Build GRCh38
rs758683600 Chr.1: 46689558-46689558 on Build GRCh38
rs373335812 Chr.1: 46689683-46689683 on Build GRCh38
rs772856332 Chr.1: 46689641-46689641 on Build GRCh38
rs565415116 Chr.1: 46689608-46689608 on Build GRCh38
rs746732151 Chr.1: 46689644-46689644 on Build GRCh38
rs992444848 Chr.1: 46689568-46689568 on Build GRCh38
rs150658610 Chr.1: 46689592-46689592 on Build GRCh38
rs376921456 Chr.1: 46689557-46689557 on Build GRCh38
rs763934098 Chr.1: 46689713-46689713 on Build GRCh38
rs759493629 Chr.1: 46689688-46689688 on Build GRCh38
rs17102441 Chr.1: 46689700-46689700 on Build GRCh38
rs771708802 Chr.1: 46689640-46689640 on Build GRCh38
rs546757407 Chr.1: 46689569-46689569 on Build GRCh38
rs756969890 Chr.1: 46689667-46689668 on Build GRCh38
rs373594474 Chr.1: 46689645-46689645 on Build GRCh38

View Full Product Details