Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 63186336-63186609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMJD1C
Entrez Gene ID
221037
Gene Name
jumonji domain containing 1C
Gene Aliases
TRIP-8, TRIP8
UniGene
Hs.413416
Gene Chromosome Location
Chr.10: 63167221-63522075 on Build GRCh38
Transcripts
NM_001282948.1 NM_001318153.1 NM_001318154.1 NM_001322252.1 NM_001322254.1 NM_001322258.1 NM_032776.2 NR_134512.1 XM_011539503.1 XM_017015897.1 XM_017015898.1 XM_017015899.1 XM_017015900.1 XM_017015901.1 XM_017015902.1 XM_017015903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
465839 c.5866G>A p.V1956M Chr.10: 63186377-63186377 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs540765795 Chr.10: 63186431-63186433 on Build GRCh38
rs35278719 Chr.10: 63186542-63186543 on Build GRCh38
rs762528604 Chr.10: 63186431-63186434 on Build GRCh38
rs763592727 Chr.10: 63186391-63186394 on Build GRCh38
rs983510485 Chr.10: 63186375-63186375 on Build GRCh38
rs562842039 Chr.10: 63186434-63186441 on Build GRCh38
rs1027389811 Chr.10: 63186536-63186536 on Build GRCh38
rs965839076 Chr.10: 63186552-63186552 on Build GRCh38
rs371203235 Chr.10: 63186392-63186392 on Build GRCh38
rs773772219 Chr.10: 63186379-63186379 on Build GRCh38
rs753779974 Chr.10: 63186416-63186416 on Build GRCh38
rs1035092326 Chr.10: 63186571-63186571 on Build GRCh38
rs750121867 Chr.10: 63186429-63186430 on Build GRCh38
rs371310737 Chr.10: 63186454-63186457 on Build GRCh38
rs572628666 Chr.10: 63186537-63186537 on Build GRCh38
rs912438462 Chr.10: 63186540-63186540 on Build GRCh38
rs1002658363 Chr.10: 63186455-63186455 on Build GRCh38
rs778662959 Chr.10: 63186422-63186422 on Build GRCh38
rs1007544573 Chr.10: 63186457-63186457 on Build GRCh38
rs1026873661 Chr.10: 63186524-63186524 on Build GRCh38
rs375459573 Chr.10: 63186398-63186398 on Build GRCh38
rs138979310 Chr.10: 63186499-63186499 on Build GRCh38
rs1004593055 Chr.10: 63186561-63186561 on Build GRCh38
rs906943181 Chr.10: 63186433-63186433 on Build GRCh38
rs527492128 Chr.10: 63186441-63186442 on Build GRCh38
rs753183943 Chr.10: 63186434-63186437 on Build GRCh38
rs750110684 Chr.10: 63186428-63186430 on Build GRCh38
rs150264825 Chr.10: 63186420-63186420 on Build GRCh38
rs763263818 Chr.10: 63186380-63186380 on Build GRCh38
rs761502617 Chr.10: 63186416-63186417 on Build GRCh38
rs763900925 Chr.10: 63186413-63186413 on Build GRCh38
rs916386614 Chr.10: 63186421-63186421 on Build GRCh38
rs995363104 Chr.10: 63186514-63186514 on Build GRCh38
rs751090141 Chr.10: 63186395-63186398 on Build GRCh38
rs759687905 Chr.10: 63186405-63186405 on Build GRCh38
rs770262916 Chr.10: 63186365-63186365 on Build GRCh38
rs747101992 Chr.10: 63186435-63186436 on Build GRCh38
rs7897511 Chr.10: 63186488-63186488 on Build GRCh38
rs552821035 Chr.10: 63186523-63186523 on Build GRCh38
rs951186301 Chr.10: 63186527-63186527 on Build GRCh38
rs777982937 Chr.10: 63186433-63186434 on Build GRCh38
rs367885257 Chr.10: 63186444-63186445 on Build GRCh38
rs750280605 Chr.10: 63186429-63186429 on Build GRCh38

View Full Product Details