Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 112574043-112574304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1958
Entrez Gene ID
158405
Gene Name
KIAA1958
Gene Aliases
-
UniGene
Hs.656619
Gene Chromosome Location
Chr.9: 112486968-112665311 on Build GRCh38
Transcripts
NM_001287036.1 NM_001287038.1 NM_133465.3 XM_011518311.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3653131 c.119C>T p.S40F Chr.9: 112574199-112574199 on Build GRCh38
5638572 c.154G>A p.A52T Chr.9: 112574234-112574234 on Build GRCh38
3653133 c.143G>T p.W48L Chr.9: 112574223-112574223 on Build GRCh38
5647743 c.23C>T p.S8L Chr.9: 112574103-112574103 on Build GRCh38
3170273 c.60T>C p.H20H Chr.9: 112574140-112574140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770167541 Chr.9: 112574239-112574239 on Build GRCh38
rs377509113 Chr.9: 112574190-112574190 on Build GRCh38
rs749805916 Chr.9: 112574262-112574262 on Build GRCh38
rs752507452 Chr.9: 112574165-112574165 on Build GRCh38
rs767698180 Chr.9: 112574149-112574149 on Build GRCh38
rs773669501 Chr.9: 112574245-112574245 on Build GRCh38
rs373112051 Chr.9: 112574183-112574183 on Build GRCh38
rs776970032 Chr.9: 112574100-112574100 on Build GRCh38
rs766512203 Chr.9: 112574131-112574131 on Build GRCh38
rs756651490 Chr.9: 112574193-112574193 on Build GRCh38
rs756020391 Chr.9: 112574173-112574173 on Build GRCh38
rs745431005 Chr.9: 112574210-112574210 on Build GRCh38
rs768849427 Chr.9: 112574095-112574095 on Build GRCh38
rs778502650 Chr.9: 112574204-112574204 on Build GRCh38
rs972853920 Chr.9: 112574263-112574263 on Build GRCh38
rs565100542 Chr.9: 112574137-112574137 on Build GRCh38
rs745535081 Chr.9: 112574081-112574081 on Build GRCh38
rs748491527 Chr.9: 112574226-112574226 on Build GRCh38
rs368680529 Chr.9: 112574185-112574185 on Build GRCh38
rs758068064 Chr.9: 112574217-112574217 on Build GRCh38
rs28580102 Chr.9: 112574203-112574203 on Build GRCh38
rs933721230 Chr.9: 112574259-112574259 on Build GRCh38
rs867260044 Chr.9: 112574103-112574103 on Build GRCh38
rs770257330 Chr.9: 112574116-112574116 on Build GRCh38
rs778385560 Chr.9: 112574068-112574068 on Build GRCh38
rs774379497 Chr.9: 112574135-112574135 on Build GRCh38
rs531258413 Chr.9: 112574282-112574282 on Build GRCh38
rs762973436 Chr.9: 112574125-112574125 on Build GRCh38
rs368954378 Chr.9: 112574076-112574076 on Build GRCh38
rs773759274 Chr.9: 112574123-112574123 on Build GRCh38
rs748581392 Chr.9: 112574112-112574112 on Build GRCh38
rs370659943 Chr.9: 112574219-112574219 on Build GRCh38

View Full Product Details