Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 20369480-20369713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSM1
Entrez Gene ID
3642
Gene Name
INSM transcriptional repressor 1
Gene Aliases
IA-1, IA1
UniGene
Hs.89584
Gene Chromosome Location
Chr.20: 20368121-20370949 on Build GRCh38
Transcripts
NM_002196.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3544571 c.1407C>T p.F469F Chr.20: 20369674-20369674 on Build GRCh38
5967559 c.1237delG p.D415fs*20 Chr.20: 20369504-20369504 on Build GRCh38
4439705 c.1390C>A p.H464N Chr.20: 20369657-20369657 on Build GRCh38
6845969 c.1413C>G p.C471W Chr.20: 20369680-20369680 on Build GRCh38
5857484 c.1408C>T p.P470S Chr.20: 20369675-20369675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745505871 Chr.20: 20369658-20369658 on Build GRCh38
rs531505955 Chr.20: 20369662-20369662 on Build GRCh38
rs748646966 Chr.20: 20369598-20369598 on Build GRCh38
rs745868622 Chr.20: 20369508-20369508 on Build GRCh38
rs765263425 Chr.20: 20369565-20369565 on Build GRCh38
rs778213243 Chr.20: 20369637-20369637 on Build GRCh38
rs746077325 Chr.20: 20369526-20369526 on Build GRCh38
rs749422329 Chr.20: 20369577-20369577 on Build GRCh38
rs770648671 Chr.20: 20369608-20369608 on Build GRCh38
rs373203712 Chr.20: 20369566-20369566 on Build GRCh38
rs970478715 Chr.20: 20369641-20369641 on Build GRCh38
rs762213940 Chr.20: 20369551-20369551 on Build GRCh38
rs753992828 Chr.20: 20369631-20369631 on Build GRCh38
rs774405537 Chr.20: 20369695-20369695 on Build GRCh38
rs778667570 Chr.20: 20369504-20369504 on Build GRCh38
rs907734322 Chr.20: 20369614-20369614 on Build GRCh38
rs752571863 Chr.20: 20369506-20369506 on Build GRCh38
rs1001196034 Chr.20: 20369578-20369578 on Build GRCh38
rs749274733 Chr.20: 20369532-20369532 on Build GRCh38
rs775638830 Chr.20: 20369618-20369618 on Build GRCh38
rs200889143 Chr.20: 20369629-20369629 on Build GRCh38
rs769233282 Chr.20: 20369543-20369543 on Build GRCh38
rs1053675785 Chr.20: 20369585-20369585 on Build GRCh38
rs771181787 Chr.20: 20369650-20369650 on Build GRCh38
rs780192370 Chr.20: 20369530-20369530 on Build GRCh38
rs779530790 Chr.20: 20369596-20369596 on Build GRCh38
rs747327681 Chr.20: 20369606-20369606 on Build GRCh38
rs759139506 Chr.20: 20369610-20369610 on Build GRCh38
rs754959370 Chr.20: 20369500-20369500 on Build GRCh38
rs564197812 Chr.20: 20369630-20369630 on Build GRCh38
rs944093350 Chr.20: 20369521-20369521 on Build GRCh38
rs774995604 Chr.20: 20369690-20369690 on Build GRCh38
rs138934668 Chr.20: 20369635-20369635 on Build GRCh38
rs574942900 Chr.20: 20369568-20369568 on Build GRCh38
rs775159800 Chr.20: 20369664-20369664 on Build GRCh38
rs755151592 Chr.20: 20369633-20369633 on Build GRCh38
rs758929654 Chr.20: 20369564-20369564 on Build GRCh38
rs760326605 Chr.20: 20369554-20369554 on Build GRCh38
rs185120151 Chr.20: 20369569-20369569 on Build GRCh38
rs756379486 Chr.20: 20369572-20369572 on Build GRCh38
rs768749964 Chr.20: 20369676-20369676 on Build GRCh38
rs765891839 Chr.20: 20369556-20369556 on Build GRCh38
rs572384565 Chr.20: 20369626-20369626 on Build GRCh38
rs773581454 Chr.20: 20369605-20369605 on Build GRCh38
rs556795808 Chr.20: 20369563-20369563 on Build GRCh38
rs774689301 Chr.20: 20369550-20369550 on Build GRCh38
rs1051936496 Chr.20: 20369649-20369649 on Build GRCh38
rs963034856 Chr.20: 20369660-20369660 on Build GRCh38
rs764643353 Chr.20: 20369611-20369611 on Build GRCh38
rs758240556 Chr.20: 20369507-20369507 on Build GRCh38
rs780094822 Chr.20: 20369576-20369576 on Build GRCh38
rs770072780 Chr.20: 20369528-20369528 on Build GRCh38
rs751338296 Chr.20: 20369571-20369571 on Build GRCh38
rs762509361 Chr.20: 20369675-20369675 on Build GRCh38
rs776407227 Chr.20: 20369653-20369653 on Build GRCh38
rs758929075 Chr.20: 20369636-20369636 on Build GRCh38
rs762979020 Chr.20: 20369533-20369550 on Build GRCh38
rs202019561 Chr.20: 20369590-20369590 on Build GRCh38
rs1023628563 Chr.20: 20369514-20369514 on Build GRCh38
rs1004580925 Chr.20: 20369648-20369648 on Build GRCh38
rs370307027 Chr.20: 20369552-20369552 on Build GRCh38
rs772498911 Chr.20: 20369602-20369602 on Build GRCh38

View Full Product Details