Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1790912-1791179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFALS
Entrez Gene ID
3483
Gene Name
insulin like growth factor binding protein acid labile subunit
Gene Aliases
ACLSD, ALS
UniGene
Hs.839
Gene Chromosome Location
Chr.16: 1790413-1794908 on Build GRCh38
Transcripts
NM_001146006.1 NM_004970.2 NR_027389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6684302 c.1513C>T p.R505C Chr.16: 1791019-1791019 on Build GRCh38
967677 c.1340C>T p.T447M Chr.16: 1791078-1791078 on Build GRCh38
6684304 c.1371C>T p.D457D Chr.16: 1791161-1791161 on Build GRCh38
5677701 c.1603T>A p.L535M Chr.16: 1790929-1790929 on Build GRCh38
398944 c.1445A>T p.Q482L Chr.16: 1790973-1790973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776840046 Chr.16: 1791100-1791100 on Build GRCh38
rs368227180 Chr.16: 1790973-1790973 on Build GRCh38
rs774754339 Chr.16: 1790997-1790997 on Build GRCh38
rs933154166 Chr.16: 1791109-1791109 on Build GRCh38
rs747262165 Chr.16: 1791079-1791079 on Build GRCh38
rs971877665 Chr.16: 1791087-1791087 on Build GRCh38
rs767214247 Chr.16: 1791139-1791139 on Build GRCh38
rs755775132 Chr.16: 1791127-1791127 on Build GRCh38
rs777354303 Chr.16: 1791135-1791135 on Build GRCh38
rs769032694 Chr.16: 1790943-1790943 on Build GRCh38
rs765159127 Chr.16: 1791116-1791116 on Build GRCh38
rs767680008 Chr.16: 1790949-1790949 on Build GRCh38
rs908610548 Chr.16: 1790966-1790966 on Build GRCh38
rs755584337 Chr.16: 1791140-1791140 on Build GRCh38
rs35393265 Chr.16: 1790959-1790959 on Build GRCh38
rs753836798 Chr.16: 1790970-1790970 on Build GRCh38
rs779354727 Chr.16: 1791074-1791074 on Build GRCh38
rs749827761 Chr.16: 1790994-1790994 on Build GRCh38
rs370640507 Chr.16: 1791093-1791093 on Build GRCh38
rs770443219 Chr.16: 1790990-1790990 on Build GRCh38
rs916659112 Chr.16: 1791080-1791080 on Build GRCh38
rs752485607 Chr.16: 1790979-1790979 on Build GRCh38
rs778249405 Chr.16: 1791157-1791157 on Build GRCh38
rs202078372 Chr.16: 1791063-1791063 on Build GRCh38
rs368802110 Chr.16: 1790996-1790996 on Build GRCh38
rs759604566 Chr.16: 1791101-1791101 on Build GRCh38
rs548958500 Chr.16: 1791012-1791012 on Build GRCh38
rs1053975229 Chr.16: 1791003-1791003 on Build GRCh38
rs756878975 Chr.16: 1791137-1791137 on Build GRCh38
rs773569527 Chr.16: 1791013-1791013 on Build GRCh38
rs780890947 Chr.16: 1790983-1790983 on Build GRCh38
rs36068954 Chr.16: 1791152-1791152 on Build GRCh38
rs762140953 Chr.16: 1791004-1791004 on Build GRCh38
rs17559 Chr.16: 1791032-1791032 on Build GRCh38
rs886051776 Chr.16: 1790952-1790952 on Build GRCh38
rs372565928 Chr.16: 1790953-1790953 on Build GRCh38
rs200380381 Chr.16: 1790940-1790940 on Build GRCh38
rs902480401 Chr.16: 1791095-1791095 on Build GRCh38
rs753116894 Chr.16: 1791158-1791158 on Build GRCh38
rs763827670 Chr.16: 1791133-1791133 on Build GRCh38
rs778796934 Chr.16: 1790974-1790974 on Build GRCh38
rs772332590 Chr.16: 1791077-1791077 on Build GRCh38
rs373710655 Chr.16: 1791019-1791019 on Build GRCh38
rs1051343231 Chr.16: 1791131-1791131 on Build GRCh38
rs746584286 Chr.16: 1790933-1790933 on Build GRCh38
rs199573954 Chr.16: 1791148-1791148 on Build GRCh38
rs745430262 Chr.16: 1791070-1791070 on Build GRCh38
rs182305760 Chr.16: 1790982-1790982 on Build GRCh38
rs372854299 Chr.16: 1791117-1791117 on Build GRCh38
rs745653849 Chr.16: 1791016-1791016 on Build GRCh38
rs9282729 Chr.16: 1790993-1790993 on Build GRCh38
rs755495272 Chr.16: 1791072-1791072 on Build GRCh38
rs752536799 Chr.16: 1791051-1791051 on Build GRCh38
rs551584413 Chr.16: 1791064-1791064 on Build GRCh38
rs762760955 Chr.16: 1791128-1791128 on Build GRCh38
rs762062477 Chr.16: 1790947-1790947 on Build GRCh38
rs751397977 Chr.16: 1791136-1791136 on Build GRCh38
rs780650874 Chr.16: 1791067-1791067 on Build GRCh38
rs149707428 Chr.16: 1791061-1791061 on Build GRCh38
rs940549283 Chr.16: 1790971-1790971 on Build GRCh38
rs564251650 Chr.16: 1790956-1790956 on Build GRCh38
rs200564045 Chr.16: 1791151-1791151 on Build GRCh38
rs376821550 Chr.16: 1791007-1791007 on Build GRCh38
rs113804102 Chr.16: 1790957-1790957 on Build GRCh38
rs765148560 Chr.16: 1791044-1791044 on Build GRCh38
rs774398035 Chr.16: 1790945-1790945 on Build GRCh38
rs369848154 Chr.16: 1791018-1791018 on Build GRCh38
rs753784566 Chr.16: 1791023-1791023 on Build GRCh38
rs544452737 Chr.16: 1790941-1790941 on Build GRCh38
rs1036128809 Chr.16: 1790978-1790978 on Build GRCh38
rs1055208203 Chr.16: 1790934-1790935 on Build GRCh38
rs758059640 Chr.16: 1790981-1790981 on Build GRCh38
rs571715026 Chr.16: 1791078-1791078 on Build GRCh38

View Full Product Details