Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62200940-62201165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTG2
Entrez Gene ID
26164
Gene Name
mitochondrial ribosome associated GTPase 2
Gene Aliases
GTPBP5, ObgH1, dJ1005F21.2
UniGene
Hs.340636
Gene Chromosome Location
Chr.20: 62183025-62203568 on Build GRCh38
Transcripts
NM_015666.3 XM_005260393.1 XM_006723774.3 XM_011528776.1 XM_017027805.1 XM_017027806.1 XM_017027807.1 XM_017027808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6716333 c.1091G>A p.G364E Chr.20: 62200947-62200947 on Build GRCh38
5583598 c.1184C>T p.A395V Chr.20: 62201040-62201040 on Build GRCh38
2766315 c.1170C>T p.D390D Chr.20: 62201026-62201026 on Build GRCh38
6716331 c.1144C>T p.L382L Chr.20: 62201000-62201000 on Build GRCh38
7143154 c.1135G>C p.E379Q Chr.20: 62200991-62200991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761253594 Chr.20: 62201018-62201018 on Build GRCh38
rs759425297 Chr.20: 62201034-62201034 on Build GRCh38
rs765177017 Chr.20: 62201106-62201106 on Build GRCh38
rs766129154 Chr.20: 62201107-62201110 on Build GRCh38
rs759302451 Chr.20: 62201093-62201093 on Build GRCh38
rs750600172 Chr.20: 62201042-62201042 on Build GRCh38
rs766731411 Chr.20: 62201121-62201121 on Build GRCh38
rs760726538 Chr.20: 62201104-62201104 on Build GRCh38
rs777760321 Chr.20: 62200968-62200968 on Build GRCh38
rs377220160 Chr.20: 62200969-62200969 on Build GRCh38
rs1004027307 Chr.20: 62201136-62201136 on Build GRCh38
rs778978065 Chr.20: 62200972-62200972 on Build GRCh38
rs572424775 Chr.20: 62201020-62201020 on Build GRCh38
rs780500068 Chr.20: 62201057-62201057 on Build GRCh38
rs748754740 Chr.20: 62201065-62201065 on Build GRCh38
rs146888379 Chr.20: 62201026-62201026 on Build GRCh38
rs758710566 Chr.20: 62201044-62201044 on Build GRCh38
rs757250378 Chr.20: 62201041-62201041 on Build GRCh38
rs560078782 Chr.20: 62200997-62200997 on Build GRCh38
rs532198011 Chr.20: 62201090-62201090 on Build GRCh38
rs781662967 Chr.20: 62201127-62201127 on Build GRCh38
rs767337108 Chr.20: 62201095-62201095 on Build GRCh38
rs985645630 Chr.20: 62201133-62201133 on Build GRCh38
rs1054885 Chr.20: 62201126-62201126 on Build GRCh38
rs547100416 Chr.20: 62201107-62201107 on Build GRCh38
rs112442787 Chr.20: 62201005-62201005 on Build GRCh38
rs897677276 Chr.20: 62201046-62201046 on Build GRCh38
rs762445306 Chr.20: 62201089-62201089 on Build GRCh38
rs368976452 Chr.20: 62200979-62200979 on Build GRCh38
rs748544788 Chr.20: 62201128-62201128 on Build GRCh38
rs912844200 Chr.20: 62200960-62200960 on Build GRCh38
rs767461500 Chr.20: 62201035-62201035 on Build GRCh38
rs780626811 Chr.20: 62200985-62200985 on Build GRCh38
rs772792354 Chr.20: 62201084-62201084 on Build GRCh38
rs750878221 Chr.20: 62201037-62201038 on Build GRCh38
rs118176146 Chr.20: 62201032-62201032 on Build GRCh38
rs752575356 Chr.20: 62201096-62201096 on Build GRCh38
rs766226812 Chr.20: 62200960-62200960 on Build GRCh38
rs754921359 Chr.20: 62200963-62200963 on Build GRCh38
rs926210125 Chr.20: 62201053-62201053 on Build GRCh38
rs772563146 Chr.20: 62200981-62200981 on Build GRCh38
rs751881753 Chr.20: 62201122-62201122 on Build GRCh38
rs143916641 Chr.20: 62200971-62200971 on Build GRCh38
rs549879158 Chr.20: 62201033-62201033 on Build GRCh38
rs200118420 Chr.20: 62201058-62201058 on Build GRCh38
rs140539598 Chr.20: 62201027-62201027 on Build GRCh38
rs769172438 Chr.20: 62201082-62201082 on Build GRCh38
rs781529784 Chr.20: 62201064-62201064 on Build GRCh38
rs143630884 Chr.20: 62201055-62201056 on Build GRCh38
rs758655096 Chr.20: 62201113-62201113 on Build GRCh38
rs776242026 Chr.20: 62201006-62201006 on Build GRCh38

View Full Product Details