Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83112700-83112973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDGFRP3
Entrez Gene ID
50810
Gene Name
hepatoma-derived growth factor, related protein 3
Gene Aliases
CGI-142, HDGF-2, HDGF2, HRP-3
UniGene
Hs.513954
Gene Chromosome Location
Chr.15: 83112738-83208018 on Build GRCh38
Transcripts
XM_006720554.3 NM_001144903.1 NM_023003.3 XM_011521679.2 XM_011521680.2 XM_017022320.1 XM_017022321.1 XM_017022322.1 XM_017022323.1 XM_017022324.1 XM_017022325.1 XM_017022326.1 XM_017022327.1 XM_017022328.1 XM_017022329.1 XM_017022330.1 XM_017022331.1

Gene Symbol
LOC105370931
Entrez Gene ID
105370931
Gene Name
uncharacterized LOC105370931
Gene Aliases
-
UniGene
Hs.667183
Gene Chromosome Location
Chr.15: 83096605-83114828 on Build GRCh38
Transcripts
XM_006720554.3 NM_001144903.1 NM_023003.3 XM_011521679.2 XM_011521680.2 XM_017022320.1 XM_017022321.1 XM_017022322.1 XM_017022323.1 XM_017022324.1 XM_017022325.1 XM_017022326.1 XM_017022327.1 XM_017022328.1 XM_017022329.1 XM_017022330.1 XM_017022331.1

Gene Symbol
TM6SF1
Entrez Gene ID
53346
Gene Name
transmembrane 6 superfamily member 1
Gene Aliases
-
UniGene
Hs.513094
Gene Chromosome Location
Chr.15: 83107572-83145403 on Build GRCh38
Transcripts
XM_006720554.3 NM_001144903.1 NM_023003.3 XM_011521679.2 XM_011521680.2 XM_017022320.1 XM_017022321.1 XM_017022322.1 XM_017022323.1 XM_017022324.1 XM_017022325.1 XM_017022326.1 XM_017022327.1 XM_017022328.1 XM_017022329.1 XM_017022330.1 XM_017022331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2012764 c.160G>A p.V54I Chr.15: 83112864-83112864 on Build GRCh38
5769808 c.127C>T p.L43L Chr.15: 83112831-83112831 on Build GRCh38
5769807 c.127C>T p.L43L Chr.15: 83112831-83112831 on Build GRCh38
3887616 c.183C>T p.D61D Chr.15: 83112887-83112887 on Build GRCh38
3816903 c.174A>C p.P58P Chr.15: 83112878-83112878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371581703 Chr.15: 83112882-83112882 on Build GRCh38
rs755830379 Chr.15: 83112815-83112815 on Build GRCh38
rs201104135 Chr.15: 83112883-83112883 on Build GRCh38
rs145491081 Chr.15: 83112864-83112864 on Build GRCh38
rs374990663 Chr.15: 83112808-83112808 on Build GRCh38
rs376049026 Chr.15: 83112784-83112784 on Build GRCh38
rs1989 Chr.15: 83112879-83112879 on Build GRCh38
rs753388918 Chr.15: 83112893-83112893 on Build GRCh38
rs374531797 Chr.15: 83112770-83112770 on Build GRCh38
rs778069710 Chr.15: 83112849-83112849 on Build GRCh38
rs564400844 Chr.15: 83112924-83112924 on Build GRCh38
rs141243867 Chr.15: 83112875-83112875 on Build GRCh38
rs747125199 Chr.15: 83112926-83112926 on Build GRCh38
rs368898788 Chr.15: 83112748-83112748 on Build GRCh38
rs368481259 Chr.15: 83112825-83112825 on Build GRCh38
rs761410387 Chr.15: 83112807-83112807 on Build GRCh38
rs771328156 Chr.15: 83112856-83112856 on Build GRCh38
rs1026387830 Chr.15: 83112746-83112746 on Build GRCh38
rs374579963 Chr.15: 83112942-83112942 on Build GRCh38
rs376138369 Chr.15: 83112904-83112904 on Build GRCh38
rs191656608 Chr.15: 83112771-83112771 on Build GRCh38
rs3803384 Chr.15: 83112863-83112863 on Build GRCh38
rs775487472 Chr.15: 83112781-83112781 on Build GRCh38
rs781500097 Chr.15: 83112934-83112934 on Build GRCh38
rs757373408 Chr.15: 83112927-83112927 on Build GRCh38
rs761856253 Chr.15: 83112797-83112807 on Build GRCh38
rs370638491 Chr.15: 83112914-83112914 on Build GRCh38
rs149678759 Chr.15: 83112855-83112855 on Build GRCh38
rs768499836 Chr.15: 83112945-83112945 on Build GRCh38
rs755264960 Chr.15: 83112833-83112833 on Build GRCh38
rs71412294 Chr.15: 83112817-83112817 on Build GRCh38
rs772235568 Chr.15: 83112768-83112768 on Build GRCh38
rs79470022 Chr.15: 83112847-83112847 on Build GRCh38
rs1075619 Chr.15: 83112722-83112722 on Build GRCh38
rs761669733 Chr.15: 83112871-83112871 on Build GRCh38
rs774118058 Chr.15: 83112793-83112793 on Build GRCh38
rs765456054 Chr.15: 83112905-83112905 on Build GRCh38
rs1012563857 Chr.15: 83112727-83112727 on Build GRCh38
rs370383601 Chr.15: 83112789-83112789 on Build GRCh38
rs1029180357 Chr.15: 83112830-83112830 on Build GRCh38
rs750080118 Chr.15: 83112813-83112813 on Build GRCh38
rs765050568 Chr.15: 83112867-83112871 on Build GRCh38
rs80329914 Chr.15: 83112846-83112846 on Build GRCh38
rs370056045 Chr.15: 83112754-83112754 on Build GRCh38
rs776467502 Chr.15: 83112764-83112764 on Build GRCh38
rs748296919 Chr.15: 83112759-83112759 on Build GRCh38
rs866607604 Chr.15: 83112887-83112887 on Build GRCh38
rs765999177 Chr.15: 83112816-83112816 on Build GRCh38
rs779502067 Chr.15: 83112845-83112845 on Build GRCh38
rs975108128 Chr.15: 83112831-83112831 on Build GRCh38

View Full Product Details