Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18435572-18435801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISYNA1
Entrez Gene ID
51477
Gene Name
inositol-3-phosphate synthase 1
Gene Aliases
INO1, INOS, IPS, IPS 1, IPS-1
UniGene
Hs.405873
Gene Chromosome Location
Chr.19: 18434388-18438301 on Build GRCh38
Transcripts
NM_001170938.1 NM_001253389.1 NM_016368.4 NR_045573.1 NR_045574.1 XM_011528059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4750665 c.1140+1G>A p.? Chr.19: 18435756-18435756 on Build GRCh38
6687658 c.1218C>T p.G406G Chr.19: 18435599-18435599 on Build GRCh38
3100051 c.1181G>A p.R394H Chr.19: 18435636-18435636 on Build GRCh38
5389125 c.1126G>A p.E376K Chr.19: 18435771-18435771 on Build GRCh38
5659997 c.1113T>C p.Y371Y Chr.19: 18435784-18435784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760638547 Chr.19: 18435746-18435746 on Build GRCh38
rs747567609 Chr.19: 18435648-18435648 on Build GRCh38
rs371089430 Chr.19: 18435707-18435707 on Build GRCh38
rs754688756 Chr.19: 18435614-18435614 on Build GRCh38
rs534754250 Chr.19: 18435745-18435745 on Build GRCh38
rs777925261 Chr.19: 18435590-18435593 on Build GRCh38
rs771257185 Chr.19: 18435655-18435655 on Build GRCh38
rs368894363 Chr.19: 18435734-18435734 on Build GRCh38
rs778230100 Chr.19: 18435629-18435629 on Build GRCh38
rs150073676 Chr.19: 18435777-18435777 on Build GRCh38
rs770066301 Chr.19: 18435671-18435671 on Build GRCh38
rs777309048 Chr.19: 18435727-18435727 on Build GRCh38
rs775764291 Chr.19: 18435739-18435739 on Build GRCh38
rs370991035 Chr.19: 18435738-18435738 on Build GRCh38
rs746131845 Chr.19: 18435742-18435743 on Build GRCh38
rs752350505 Chr.19: 18435722-18435722 on Build GRCh38
rs753681814 Chr.19: 18435714-18435714 on Build GRCh38
rs768823788 Chr.19: 18435744-18435744 on Build GRCh38
rs760734355 Chr.19: 18435705-18435705 on Build GRCh38
rs925770815 Chr.19: 18435649-18435649 on Build GRCh38
rs900189747 Chr.19: 18435608-18435608 on Build GRCh38
rs764939854 Chr.19: 18435721-18435721 on Build GRCh38
rs778239804 Chr.19: 18435634-18435634 on Build GRCh38
rs986416585 Chr.19: 18435662-18435662 on Build GRCh38
rs780043482 Chr.19: 18435605-18435605 on Build GRCh38
rs757786383 Chr.19: 18435733-18435733 on Build GRCh38
rs777014766 Chr.19: 18435656-18435656 on Build GRCh38
rs754522121 Chr.19: 18435718-18435718 on Build GRCh38
rs775362357 Chr.19: 18435673-18435673 on Build GRCh38
rs373630499 Chr.19: 18435599-18435599 on Build GRCh38
rs1041004229 Chr.19: 18435725-18435725 on Build GRCh38
rs761595027 Chr.19: 18435592-18435592 on Build GRCh38
rs746030986 Chr.19: 18435663-18435663 on Build GRCh38
rs781513874 Chr.19: 18435729-18435734 on Build GRCh38
rs753399223 Chr.19: 18435611-18435611 on Build GRCh38
rs948701482 Chr.19: 18435749-18435749 on Build GRCh38
rs749570549 Chr.19: 18435740-18435740 on Build GRCh38
rs879283286 Chr.19: 18435781-18435781 on Build GRCh38
rs757165724 Chr.19: 18435728-18435729 on Build GRCh38
rs755959374 Chr.19: 18435604-18435604 on Build GRCh38
rs1022172063 Chr.19: 18435623-18435623 on Build GRCh38
rs751368711 Chr.19: 18435712-18435713 on Build GRCh38
rs1010495809 Chr.19: 18435620-18435620 on Build GRCh38
rs773261377 Chr.19: 18435700-18435700 on Build GRCh38
rs373667789 Chr.19: 18435728-18435728 on Build GRCh38
rs192195701 Chr.19: 18435682-18435682 on Build GRCh38
rs747699372 Chr.19: 18435632-18435632 on Build GRCh38
rs978027437 Chr.19: 18435719-18435719 on Build GRCh38
rs746244619 Chr.19: 18435735-18435735 on Build GRCh38
rs757886478 Chr.19: 18435723-18435723 on Build GRCh38
rs545910008 Chr.19: 18435778-18435778 on Build GRCh38
rs759362993 Chr.19: 18435775-18435775 on Build GRCh38
rs776501176 Chr.19: 18435774-18435774 on Build GRCh38

View Full Product Details