Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26458376-26458649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPS4
Entrez Gene ID
89781
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases
BLOC3S2, LE
UniGene
Hs.474436
Gene Chromosome Location
Chr.22: 26443613-26483863 on Build GRCh38
Transcripts
NM_022081.5 NM_152841.2 NR_073135.1 NR_073136.1 XM_006724353.3 XM_006724354.3 XM_006724360.3 XM_011530485.1 XM_011530486.2 XM_011530487.2 XM_011530488.2 XM_011530489.2 XM_011530490.2 XM_011530491.2 XM_011530492.1 XM_011530493.2 XM_011530494.2 XM_011530495.2 XM_011530496.2 XM_017029042.1 XM_017029043.1 XM_017029044.1 XM_017029045.1 XM_017029046.1 XM_017029047.1 XM_017029048.1 XM_017029049.1 XM_017029050.1 XM_017029051.1 XM_017029052.1 XM_017029053.1 XM_017029054.1 XM_017029055.1 XM_017029056.1 XM_017029057.1 XM_017029058.1 XM_017029059.1 XM_017029060.1 XM_017029061.1 XM_017029062.1 XM_017029063.1 XM_017029064.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1740739 c.1718A>G p.H573R Chr.22: 26458573-26458573 on Build GRCh38
4531765 c.1822G>A p.D608N Chr.22: 26458469-26458469 on Build GRCh38
6526431 c.1783G>T p.A595S Chr.22: 26458508-26458508 on Build GRCh38
3842362 c.1794G>A p.T598T Chr.22: 26458497-26458497 on Build GRCh38
4103177 c.1754A>G p.Y585C Chr.22: 26458576-26458576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983626889 Chr.22: 26458515-26458515 on Build GRCh38
rs781375770 Chr.22: 26458465-26458465 on Build GRCh38
rs764040685 Chr.22: 26458597-26458597 on Build GRCh38
rs541601069 Chr.22: 26458524-26458524 on Build GRCh38
rs772632659 Chr.22: 26458489-26458489 on Build GRCh38
rs765161412 Chr.22: 26458513-26458513 on Build GRCh38
rs200607238 Chr.22: 26458415-26458415 on Build GRCh38
rs776169421 Chr.22: 26458504-26458504 on Build GRCh38
rs145481980 Chr.22: 26458544-26458544 on Build GRCh38
rs756278524 Chr.22: 26458535-26458535 on Build GRCh38
rs371342669 Chr.22: 26458522-26458522 on Build GRCh38
rs745665820 Chr.22: 26458470-26458471 on Build GRCh38
rs779303648 Chr.22: 26458480-26458480 on Build GRCh38
rs758970516 Chr.22: 26458506-26458506 on Build GRCh38
rs770643481 Chr.22: 26458585-26458585 on Build GRCh38
rs772546709 Chr.22: 26458560-26458560 on Build GRCh38
rs748643364 Chr.22: 26458557-26458557 on Build GRCh38
rs1045376952 Chr.22: 26458556-26458556 on Build GRCh38
rs759070239 Chr.22: 26458587-26458587 on Build GRCh38
rs375744173 Chr.22: 26458418-26458418 on Build GRCh38
rs374277298 Chr.22: 26458593-26458593 on Build GRCh38
rs778239048 Chr.22: 26458562-26458562 on Build GRCh38
rs766612334 Chr.22: 26458609-26458609 on Build GRCh38
rs374834354 Chr.22: 26458413-26458413 on Build GRCh38
rs758138934 Chr.22: 26458455-26458455 on Build GRCh38
rs751313927 Chr.22: 26458606-26458606 on Build GRCh38
rs775972251 Chr.22: 26458412-26458412 on Build GRCh38
rs747411315 Chr.22: 26458564-26458564 on Build GRCh38
rs776157678 Chr.22: 26458586-26458586 on Build GRCh38
rs999611252 Chr.22: 26458474-26458474 on Build GRCh38
rs769250305 Chr.22: 26458588-26458588 on Build GRCh38
rs754076687 Chr.22: 26458610-26458610 on Build GRCh38
rs372420536 Chr.22: 26458417-26458417 on Build GRCh38
rs773058129 Chr.22: 26458420-26458420 on Build GRCh38
rs761599881 Chr.22: 26458607-26458607 on Build GRCh38
rs779471156 Chr.22: 26458552-26458552 on Build GRCh38
rs565596162 Chr.22: 26458495-26458495 on Build GRCh38
rs982508737 Chr.22: 26458551-26458551 on Build GRCh38
rs754899131 Chr.22: 26458437-26458437 on Build GRCh38
rs775463811 Chr.22: 26458591-26458591 on Build GRCh38
rs752467533 Chr.22: 26458454-26458454 on Build GRCh38
rs760258124 Chr.22: 26458432-26458432 on Build GRCh38
rs750621357 Chr.22: 26458532-26458532 on Build GRCh38
rs769405747 Chr.22: 26458472-26458473 on Build GRCh38
rs797045615 Chr.22: 26458576-26458576 on Build GRCh38
rs780284817 Chr.22: 26458539-26458539 on Build GRCh38
rs746151193 Chr.22: 26458466-26458466 on Build GRCh38
rs368334509 Chr.22: 26458477-26458477 on Build GRCh38
rs767141806 Chr.22: 26458419-26458419 on Build GRCh38
rs756155749 Chr.22: 26458470-26458470 on Build GRCh38
rs1894706 Chr.22: 26458475-26458475 on Build GRCh38
rs779927531 Chr.22: 26458552-26458552 on Build GRCh38
rs149689335 Chr.22: 26458498-26458498 on Build GRCh38
rs768878482 Chr.22: 26458414-26458414 on Build GRCh38
rs748533902 Chr.22: 26458483-26458483 on Build GRCh38
rs770733264 Chr.22: 26458503-26458503 on Build GRCh38
rs765121677 Chr.22: 26458450-26458450 on Build GRCh38
rs370380305 Chr.22: 26458433-26458433 on Build GRCh38
rs34142623 Chr.22: 26458431-26458432 on Build GRCh38
rs749347544 Chr.22: 26458476-26458476 on Build GRCh38
rs763915972 Chr.22: 26458525-26458525 on Build GRCh38
rs755073092 Chr.22: 26458549-26458549 on Build GRCh38

View Full Product Details