Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160300776-160301001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT80
Entrez Gene ID
57560
Gene Name
intraflagellar transport 80
Gene Aliases
ATD2, SRTD2, WDR56
UniGene
Hs.478095
Gene Chromosome Location
Chr.3: 160256986-160399532 on Build GRCh38
Transcripts
NM_001190241.1 NM_001190242.1 NM_020800.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6587892 c.1303G>C p.A435P Chr.3: 160300895-160300895 on Build GRCh38
4970838 c.1306G>A p.D436N Chr.3: 160300892-160300892 on Build GRCh38
5414903 c.1315+10T>A p.? Chr.3: 160300873-160300873 on Build GRCh38
4114902 c.1205G>A p.R402H Chr.3: 160300993-160300993 on Build GRCh38
5962299 c.1315+9A>T p.? Chr.3: 160300874-160300874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971761468 Chr.3: 160300866-160300866 on Build GRCh38
rs765339125 Chr.3: 160300875-160300875 on Build GRCh38
rs1016473050 Chr.3: 160300835-160300835 on Build GRCh38
rs761280914 Chr.3: 160300836-160300836 on Build GRCh38
rs749233164 Chr.3: 160300914-160300914 on Build GRCh38
rs764632130 Chr.3: 160300839-160300839 on Build GRCh38
rs918952642 Chr.3: 160300891-160300891 on Build GRCh38
rs770394750 Chr.3: 160300915-160300915 on Build GRCh38
rs781578119 Chr.3: 160300917-160300919 on Build GRCh38
rs775951558 Chr.3: 160300833-160300833 on Build GRCh38
rs9637414 Chr.3: 160300854-160300854 on Build GRCh38
rs985991821 Chr.3: 160300827-160300827 on Build GRCh38
rs758606810 Chr.3: 160300887-160300887 on Build GRCh38
rs777754118 Chr.3: 160300906-160300906 on Build GRCh38
rs750652226 Chr.3: 160300883-160300883 on Build GRCh38
rs112636069 Chr.3: 160300823-160300823 on Build GRCh38
rs754395758 Chr.3: 160300852-160300852 on Build GRCh38
rs748511587 Chr.3: 160300943-160300944 on Build GRCh38
rs747879864 Chr.3: 160300895-160300895 on Build GRCh38
rs756047692 Chr.3: 160300896-160300896 on Build GRCh38
rs780986098 Chr.3: 160300889-160300889 on Build GRCh38
rs773877185 Chr.3: 160300930-160300930 on Build GRCh38
rs745534739 Chr.3: 160300955-160300955 on Build GRCh38
rs755456639 Chr.3: 160300865-160300866 on Build GRCh38
rs768030857 Chr.3: 160300832-160300832 on Build GRCh38
rs1003489054 Chr.3: 160300831-160300831 on Build GRCh38
rs961664932 Chr.3: 160300838-160300840 on Build GRCh38
rs527756323 Chr.3: 160300901-160300901 on Build GRCh38
rs776006563 Chr.3: 160300976-160300976 on Build GRCh38
rs371535660 Chr.3: 160300850-160300850 on Build GRCh38
rs148396265 Chr.3: 160300969-160300969 on Build GRCh38
rs150087133 Chr.3: 160300813-160300813 on Build GRCh38

View Full Product Details