Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45004107-45004354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECTD3
Entrez Gene ID
79654
Gene Name
HECT domain E3 ubiquitin protein ligase 3
Gene Aliases
-
UniGene
Hs.525084
Gene Chromosome Location
Chr.1: 45002548-45011355 on Build GRCh38
Transcripts
NM_024602.5 XM_011542140.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
910109 c.2289C>A p.F763L Chr.1: 45004118-45004118 on Build GRCh38
910106 c.2289C>T p.F763F Chr.1: 45004118-45004118 on Build GRCh38
5571814 c.2178A>G p.V726V Chr.1: 45004342-45004342 on Build GRCh38
5571815 c.1326A>G p.V442V Chr.1: 45004342-45004342 on Build GRCh38
4637713 c.1019T>C p.V340A Chr.1: 45004331-45004331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555998898 Chr.1: 45004176-45004176 on Build GRCh38
rs777419225 Chr.1: 45004214-45004215 on Build GRCh38
rs754164067 Chr.1: 45004197-45004197 on Build GRCh38
rs751988735 Chr.1: 45004200-45004204 on Build GRCh38
rs374634395 Chr.1: 45004183-45004183 on Build GRCh38
rs747929331 Chr.1: 45004255-45004255 on Build GRCh38
rs199825235 Chr.1: 45004141-45004141 on Build GRCh38
rs750648773 Chr.1: 45004211-45004211 on Build GRCh38
rs746910209 Chr.1: 45004128-45004128 on Build GRCh38
rs780272957 Chr.1: 45004214-45004214 on Build GRCh38
rs758587928 Chr.1: 45004212-45004212 on Build GRCh38
rs754881102 Chr.1: 45004222-45004222 on Build GRCh38
rs748936991 Chr.1: 45004257-45004257 on Build GRCh38
rs770486371 Chr.1: 45004265-45004265 on Build GRCh38
rs542274257 Chr.1: 45004173-45004173 on Build GRCh38
rs758641665 Chr.1: 45004199-45004200 on Build GRCh38
rs751602144 Chr.1: 45004219-45004219 on Build GRCh38
rs761970903 Chr.1: 45004203-45004203 on Build GRCh38
rs751621651 Chr.1: 45004335-45004335 on Build GRCh38
rs369750324 Chr.1: 45004215-45004219 on Build GRCh38
rs375646592 Chr.1: 45004300-45004300 on Build GRCh38
rs769456782 Chr.1: 45004256-45004256 on Build GRCh38
rs780884717 Chr.1: 45004245-45004245 on Build GRCh38
rs145406906 Chr.1: 45004200-45004201 on Build GRCh38
rs898755735 Chr.1: 45004327-45004327 on Build GRCh38
rs199539620 Chr.1: 45004331-45004331 on Build GRCh38
rs370039448 Chr.1: 45004316-45004316 on Build GRCh38
rs776470528 Chr.1: 45004132-45004132 on Build GRCh38
rs538414715 Chr.1: 45004328-45004328 on Build GRCh38
rs762286609 Chr.1: 45004306-45004306 on Build GRCh38
rs765505591 Chr.1: 45004205-45004205 on Build GRCh38
rs375684641 Chr.1: 45004291-45004291 on Build GRCh38
rs780950399 Chr.1: 45004155-45004155 on Build GRCh38
rs769452119 Chr.1: 45004148-45004148 on Build GRCh38
rs1021656319 Chr.1: 45004332-45004332 on Build GRCh38
rs769166045 Chr.1: 45004297-45004297 on Build GRCh38
rs768398049 Chr.1: 45004131-45004131 on Build GRCh38
rs770398564 Chr.1: 45004175-45004175 on Build GRCh38
rs868036565 Chr.1: 45004188-45004188 on Build GRCh38
rs979295953 Chr.1: 45004189-45004189 on Build GRCh38
rs766576380 Chr.1: 45004334-45004334 on Build GRCh38
rs750655696 Chr.1: 45004154-45004154 on Build GRCh38
rs773794015 Chr.1: 45004288-45004288 on Build GRCh38
rs764378114 Chr.1: 45004184-45004184 on Build GRCh38

View Full Product Details