Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52799305-52799578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH3
Entrez Gene ID
3699
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
Gene Aliases
H3P, ITI-HC3, SHAP
UniGene
Hs.76716
Gene Chromosome Location
Chr.3: 52794768-52809009 on Build GRCh38
Transcripts
NM_002217.3 XM_005265105.4 XM_017006356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
446854 c.815T>A p.F272Y Chr.3: 52799397-52799397 on Build GRCh38
4572572 c.791T>A p.I264K Chr.3: 52799373-52799373 on Build GRCh38
4119213 c.839T>C p.V280A Chr.3: 52799421-52799421 on Build GRCh38
3365266 c.846T>C p.P282P Chr.3: 52799428-52799428 on Build GRCh38
4572571 c.791T>A p.I264K Chr.3: 52799373-52799373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770475192 Chr.3: 52799458-52799458 on Build GRCh38
rs755718326 Chr.3: 52799452-52799452 on Build GRCh38
rs771772174 Chr.3: 52799333-52799333 on Build GRCh38
rs758669573 Chr.3: 52799390-52799390 on Build GRCh38
rs746665566 Chr.3: 52799529-52799529 on Build GRCh38
rs777335823 Chr.3: 52799527-52799527 on Build GRCh38
rs887573201 Chr.3: 52799383-52799383 on Build GRCh38
rs368285263 Chr.3: 52799489-52799489 on Build GRCh38
rs749704167 Chr.3: 52799437-52799437 on Build GRCh38
rs765314854 Chr.3: 52799475-52799475 on Build GRCh38
rs28463071 Chr.3: 52799524-52799524 on Build GRCh38
rs200933666 Chr.3: 52799535-52799535 on Build GRCh38
rs570674211 Chr.3: 52799427-52799427 on Build GRCh38
rs377731401 Chr.3: 52799349-52799349 on Build GRCh38
rs902415072 Chr.3: 52799379-52799379 on Build GRCh38
rs753823694 Chr.3: 52799363-52799363 on Build GRCh38
rs770546252 Chr.3: 52799530-52799530 on Build GRCh38
rs1013581705 Chr.3: 52799488-52799488 on Build GRCh38
rs752135861 Chr.3: 52799434-52799434 on Build GRCh38
rs373963037 Chr.3: 52799520-52799520 on Build GRCh38
rs769812533 Chr.3: 52799532-52799532 on Build GRCh38
rs374812334 Chr.3: 52799491-52799491 on Build GRCh38
rs370980347 Chr.3: 52799405-52799405 on Build GRCh38
rs752507403 Chr.3: 52799484-52799484 on Build GRCh38
rs751651271 Chr.3: 52799412-52799412 on Build GRCh38
rs976941463 Chr.3: 52799542-52799543 on Build GRCh38
rs922863466 Chr.3: 52799543-52799543 on Build GRCh38
rs932613943 Chr.3: 52799543-52799543 on Build GRCh38
rs757382598 Chr.3: 52799413-52799413 on Build GRCh38
rs776067680 Chr.3: 52799459-52799459 on Build GRCh38
rs765075522 Chr.3: 52799367-52799367 on Build GRCh38
rs780581349 Chr.3: 52799519-52799519 on Build GRCh38
rs1001487574 Chr.3: 52799541-52799541 on Build GRCh38
rs539289176 Chr.3: 52799435-52799435 on Build GRCh38
rs766176088 Chr.3: 52799350-52799350 on Build GRCh38
rs750530761 Chr.3: 52799503-52799503 on Build GRCh38
rs764364629 Chr.3: 52799487-52799487 on Build GRCh38
rs990493920 Chr.3: 52799495-52799495 on Build GRCh38
rs759183130 Chr.3: 52799364-52799364 on Build GRCh38
rs781494651 Chr.3: 52799426-52799426 on Build GRCh38
rs762870922 Chr.3: 52799486-52799486 on Build GRCh38
rs546752822 Chr.3: 52799474-52799474 on Build GRCh38
rs760043937 Chr.3: 52799455-52799455 on Build GRCh38
rs757217846 Chr.3: 52799500-52799500 on Build GRCh38
rs752597386 Chr.3: 52799386-52799386 on Build GRCh38
rs141881202 Chr.3: 52799511-52799511 on Build GRCh38
rs760261520 Chr.3: 52799343-52799343 on Build GRCh38
rs375241452 Chr.3: 52799453-52799453 on Build GRCh38
rs767327440 Chr.3: 52799338-52799338 on Build GRCh38

View Full Product Details