Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83309916-83310189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPSE
Entrez Gene ID
10855
Gene Name
heparanase
Gene Aliases
HPA, HPA1, HPR1, HPSE1, HSE1
UniGene
Hs.44227
Gene Chromosome Location
Chr.4: 83292461-83335153 on Build GRCh38
Transcripts
NM_001098540.2 NM_001166498.2 NM_001199830.1 NM_006665.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6681999 c.843-5delT p.? Chr.4: 83310083-83310083 on Build GRCh38
420493 c.871G>C p.D291H Chr.4: 83310050-83310050 on Build GRCh38
4691111 c.888T>C p.H296H Chr.4: 83310033-83310033 on Build GRCh38
1486125 c.843-1G>T p.? Chr.4: 83310079-83310079 on Build GRCh38
4126386 c.890A>G p.H297R Chr.4: 83310031-83310031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768557749 Chr.4: 83310106-83310106 on Build GRCh38
rs771866590 Chr.4: 83310121-83310121 on Build GRCh38
rs756263255 Chr.4: 83310081-83310081 on Build GRCh38
rs949083145 Chr.4: 83310145-83310145 on Build GRCh38
rs757650530 Chr.4: 83310055-83310055 on Build GRCh38
rs752051171 Chr.4: 83310054-83310054 on Build GRCh38
rs758142594 Chr.4: 83310118-83310119 on Build GRCh38
rs750091899 Chr.4: 83310083-83310083 on Build GRCh38
rs200672095 Chr.4: 83310091-83310091 on Build GRCh38
rs910468723 Chr.4: 83310100-83310100 on Build GRCh38
rs747977853 Chr.4: 83310115-83310115 on Build GRCh38
rs563568233 Chr.4: 83309957-83309957 on Build GRCh38
rs760265960 Chr.4: 83309988-83309988 on Build GRCh38
rs145834129 Chr.4: 83310136-83310136 on Build GRCh38
rs749834578 Chr.4: 83309985-83309985 on Build GRCh38
rs765902161 Chr.4: 83309992-83309992 on Build GRCh38
rs560761426 Chr.4: 83309982-83309982 on Build GRCh38
rs767427759 Chr.4: 83309954-83309957 on Build GRCh38
rs774229779 Chr.4: 83310109-83310109 on Build GRCh38
rs778180701 Chr.4: 83310047-83310047 on Build GRCh38
rs758732156 Chr.4: 83310034-83310034 on Build GRCh38
rs767501353 Chr.4: 83309983-83309983 on Build GRCh38
rs146047984 Chr.4: 83310075-83310075 on Build GRCh38
rs753539390 Chr.4: 83310021-83310021 on Build GRCh38
rs963130503 Chr.4: 83309999-83309999 on Build GRCh38
rs528652616 Chr.4: 83310137-83310137 on Build GRCh38
rs750921068 Chr.4: 83310090-83310090 on Build GRCh38
rs971774454 Chr.4: 83310007-83310007 on Build GRCh38
rs377096811 Chr.4: 83310094-83310094 on Build GRCh38
rs781725820 Chr.4: 83310068-83310068 on Build GRCh38

View Full Product Details