Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 134978474-134978735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBS1L
Entrez Gene ID
10767
Gene Name
HBS1 like translational GTPase
Gene Aliases
EF-1a, ERFS, HBS1, HSPC276, eRF3c
UniGene
Hs.378532
Gene Chromosome Location
Chr.6: 134960378-135054898 on Build GRCh38
Transcripts
NM_001145158.1 NM_006620.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1073593 c.1765A>C p.N589H Chr.6: 134978711-134978711 on Build GRCh38
5809498 c.1751G>T p.R584L Chr.6: 134978725-134978725 on Build GRCh38
3829108 c.1758C>T p.L586L Chr.6: 134978718-134978718 on Build GRCh38
7135286 c.1764T>C p.F588F Chr.6: 134978712-134978712 on Build GRCh38
6241092 c.1761C>A p.I587I Chr.6: 134978715-134978715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866400081 Chr.6: 134978525-134978525 on Build GRCh38
rs746744824 Chr.6: 134978648-134978648 on Build GRCh38
rs747997372 Chr.6: 134978648-134978649 on Build GRCh38
rs568990129 Chr.6: 134978519-134978519 on Build GRCh38
rs552376260 Chr.6: 134978514-134978514 on Build GRCh38
rs750643731 Chr.6: 134978634-134978634 on Build GRCh38
rs9376077 Chr.6: 134978575-134978575 on Build GRCh38
rs1008381277 Chr.6: 134978705-134978705 on Build GRCh38
rs78503088 Chr.6: 134978550-134978550 on Build GRCh38
rs748969033 Chr.6: 134978710-134978710 on Build GRCh38
rs754989690 Chr.6: 134978653-134978653 on Build GRCh38
rs918309812 Chr.6: 134978556-134978556 on Build GRCh38
rs991836178 Chr.6: 134978601-134978601 on Build GRCh38
rs909700235 Chr.6: 134978538-134978538 on Build GRCh38
rs747651830 Chr.6: 134978689-134978689 on Build GRCh38
rs948289271 Chr.6: 134978642-134978642 on Build GRCh38
rs764922640 Chr.6: 134978664-134978664 on Build GRCh38
rs760156515 Chr.6: 134978533-134978538 on Build GRCh38
rs779998392 Chr.6: 134978644-134978644 on Build GRCh38
rs533947352 Chr.6: 134978623-134978623 on Build GRCh38
rs373468324 Chr.6: 134978696-134978696 on Build GRCh38
rs935477809 Chr.6: 134978659-134978659 on Build GRCh38
rs758192329 Chr.6: 134978636-134978636 on Build GRCh38
rs776238065 Chr.6: 134978573-134978573 on Build GRCh38
rs777351501 Chr.6: 134978698-134978698 on Build GRCh38
rs10484496 Chr.6: 134978544-134978544 on Build GRCh38
rs781236257 Chr.6: 134978661-134978661 on Build GRCh38
rs775470015 Chr.6: 134978515-134978519 on Build GRCh38
rs34033376 Chr.6: 134978653-134978654 on Build GRCh38

View Full Product Details